SAB4700062
Monoclonal Anti-CD4-FITC , (N-terminal) antibody produced in mouse
clone MEM-241, purified immunoglobulin, buffered aqueous solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.44
Recommended Products
biological source
mouse
Quality Level
conjugate
FITC conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
MEM-241, monoclonal
form
buffered aqueous solution
species reactivity
human
technique(s)
flow cytometry: suitable
isotype
IgG1
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... CD4(920)
Related Categories
General description
Cluster of differentiation 4 (CD4) molecule is encoded by the gene mapped to human chromosome 12p13.31. The encoded protein is mainly localized on the surface of specific leukocytes. CD4 is characterized with a four ecto-domains (D1-D4) of which D1, D2 and D4 contain disulfide bonds.
The antibody MEM-241 recognizes CD4 antigen, a 55 kDa transmebrane glycoprotein expressed on a subset of T lymphocytes ("helper" T-cells) and also on monocytes, tissue macrophages and granulocytes.
Immunogen
2 N-terminal domains of human CD4 fused to human IgG1 Fc
Application
The reagent is designed for Flow Cytometry analysis of human blood cells using 20 μL reagent / 100 μL of whole blood or 1e6 cells in a suspension. The content of a vial (2 mL) is sufficient for 100 tests.
Biochem/physiol Actions
Cluster of differentiation 4 (CD4) facilitates the activation of immunostimulatory T-cells. It serves as a primary receptor for human immunodeficiency virus (HIV)-1 entry. The encoded protein also plays a key role in altering cell elasticity. Elevated expression of soluble CD4 has been observed in patients of infectious and chronic inflammatory diseases and rheumatoid arthritis (RA). Thus, this protein can be used as a potential diagnostic marker for the development of RA.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
Solution in phosphate buffered saline containing 15 mM sodium azide and 0.2% high-grade protease free BSA as a stabilizing agent.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Disease severity in rheumatoid arthritis: Relationships of plasma tumor necrosis factor-α, soluble interleukin 2-receptor, soluble CD4/CD8 ratio, neopterin, and fibrin D-dimer to traditional severity and functional measures
Beckham JC, et al.
Journal of clinical immunology, 12(5), 353-361 (1992)
The role of CD4 on mechanical properties of live cell membrane
Bui VC and Nguyen TH
Journal of Biomedical Materials Research Part A, 104(1), 239-244 (2016)
Isochromosomes 12p and 9p: parental origin and possible mechanisms of formation
Dutly F, et al.
European Journal of Human Genetics, 6(2), 140-144 (1998)
Human CD4 Metastability Is a Function of the Allosteric Disulfide Bond in Domain 2
Owen GR, et al.
Biochemistry, 55(15), 2227-2237 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service