Skip to Content
Merck
All Photos(2)

Key Documents

SAB4504751

Sigma-Aldrich

Anti-phospho-BCL2 (pSer70) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 26 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer70)

Gene Information

human ... BCL2(596)

Immunogen

The antiserum was produced against synthesized peptide derived from human BCL2 around the phosphorylation site of Ser70.

Immunogen Range: 36-85

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Wang et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 52, 147-156 (2019-01-03)
As an effective antimalarial medicine, Dihydroartemisinin (DHA) has therapeutic potential on human cervical cancer. However, its working mechanism has not been elucidated. This study aimed to investigate the reversal effect of DHA on human cervical cancer HeLa cells, and explored
Qiong Yu et al.
The journal of obstetrics and gynaecology research, 47(11), 3789-3796 (2021-09-11)
To investigate the optimal dose of mifepristone and lithospermum combination regimen on medical abortion in early pregnancy rats without increasing side effects. Sixty sexually mature female Sprague Dawley (SD) rats with early pregnancy were randomly allocated into 10 groups, including
Jiao Zheng et al.
Cancer management and research, 11, 5231-5242 (2019-06-27)
Background: Hepatocellular carcinoma (HCC) is a particularly prevalent type of liver cancer and is one of the deadliest malignancies in Asia. Tangeretin is a biological compound extracted from traditional Chinese herbs and has been shown to have potential antitumour properties;
Monica Nanni et al.
Cells, 8(7) (2019-07-03)
Signaling of the epithelial splice variant of fibroblast growth factor receptor 2 (FGFR2b) triggers both differentiation and autophagy, while the aberrant expression of the mesenchymal FGFR2c isoform in epithelial cells induces impaired differentiation, epithelial mesenchymal transition (EMT) and tumorigenic features.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service