Skip to Content
Merck
All Photos(3)

Key Documents

SAB4503501

Sigma-Aldrich

Anti-Cyclin B1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CCN-2, CCNB, CCNB1, CCNB1-RS1, CYCB

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 48 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CCNB1(891)

General description

Anti-Cyclin B1 Antibody detects endogenous levels of total Cyclin B1 protein.
Cyclin B1 belongs to the cyclin family. It is a 48.337kDa protein. It is present in low level in testis, thymus, bone marrow and smooth muscle. It is mapped to human chromosome 5q13.2.

Immunogen

The antiserum was produced against synthesized peptide derived from human Cyclin B1.

Immunogen Range: 91-140

Application

Anti-Cyclin B1 antibody has been used in western-blot analysis.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Cyclin B1-CDK1 (cyclin-dependent kinase 1) helps to induce chromosome condensation, nuclear lamina resolution and mitotic spindle assembly. It also acts as a potential target for a cancer vaccine.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CCNB1 (cyclin B1)
Perez-Stable C
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2009)
PIWIL2 promotes progression of non-small cell lung cancer by inducing CDK2 and Cyclin A expression
Qu X, et al.
Journal of Translational Medicine, 13(1), 301-301 (2015)
The Tumor Antigen Cyclin B1 Hosts Multiple CD4 T Cell Epitopes Differently Recognized by Pre-Existing Naive and Memory Cells in Both Healthy and Cancer Donors
Chevaleyre C, et al.
Journal of Immunology, 195(4), 1891-1901 (2015)
Identification of a novel Polo-like kinase 1 inhibitor that specifically blocks the functions of Polo-Box domain
Chen Y, et al.
Oncotarget, 8(1), 1234-1246 (2017)
Yu-Jie Wu et al.
Journal of cell science, 134(9) (2021-05-12)
Multiciliated cells (MCCs) are terminally differentiated postmitotic cells that possess hundreds of motile cilia on their apical surface. Defects in cilia formation are associated with ciliopathies that affect many organs. In this study, we tested the role and mechanism of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service