Skip to Content
Merck
All Photos(2)

Key Documents

SAB4503227

Sigma-Aldrich

Anti-OR51E2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

HPRAJ, OR51E2, Olfactory receptor 51E2, Olfactory receptor OR11-16, Prostate-specific G-protein coupled receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 35 kDa

species reactivity

rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... O51E2(81285)

Related Categories

General description

Anti-OR51E2 Antibody detects endogenous levels of total OR51E2 protein.

OR51E2 (olfactory receptor family 51 subfamily E member 2),also known as PSGR, codes for a G-protein–coupled receptor. It is a member of the odorant receptors family. OR51E2 is present in early endosomes. It is mapped to human chromosome 11p15.

Immunogen

The antiserum was produced against synthesized peptide derived from human OR51E2.

Immunogen Range: 221-270

Biochem/physiol Actions

OR51E2 (olfactory receptor family 51 subfamily E member 2) helps to mediate rapid, non-genomic, steroidal signaling in prostate cancer cells. It also promotes the growth of early prostate cancer.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Increased expression of prostate-specific G-protein-coupled receptor in human prostate intraepithelial neoplasia and prostate cancers
Weng J, et al.
International Journal of Cancer. Journal International Du Cancer, 113(5), 811-818 (2005)
Novel 5' fusion partners of ETV1 and ETV4 in prostate cancer
Barros-Silva JD, et al.
Neoplasia, 15(7), 720-726 (2013)
The Cancer Genome Atlas Clinical Explorer: a web and mobile interface for identifying clinical-genomic driver associations
Lee H, et al.
Genome Medicine, 7(1), 112-112 (2015)
Functional Characterization of the Odorant Receptor 51E2 in Human Melanocytes
Gelis L, et al.
The Journal of Biological Chemistry, 291(34), 17772-17786 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service