Skip to Content
Merck
All Photos(1)

Key Documents

SAB4503051

Sigma-Aldrich

Anti-Tyrosinase antibody produced in rabbit

affinity isolated antibody

Synonym(s):

LB24-AB, Monophenol monooxygenase, SK29-AB, TYR, Tumor rejection antigen AB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 60 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TYR(7299)

General description

Anti-Tyrosinase Antibody detects endogenous levels of total Tyrosinase protein.
The TYR (tyrosinase) gene is mapped to human chromosome 11q14.3 and codes for a membrane glycoprotein, tyrosinase. It is ubiquitously expressed in plant and animal tissue.

Immunogen

The antiserum was produced against synthesized peptide derived from human Tyrosinase.

Immunogen Range: 471-520

Biochem/physiol Actions

The encoded protein, tyrosinase, is involved in melanin production by catalyzing the oxidation of L-tyrosine and L- 3, 4-dihydroxyphenylalanine to form dopaquinone. TYR (tyrosinase) is associated with the development of neural crest cell into melanocytes. Biallelic mutations in the TYR gene results in an autosomal recessive disorder, oculocutaneous albinism type 1. Loss of tyrosinase activity contributes to the severity of this disorder, in which melanin is undetectable. Abnormal expression of the gene has been observed in atypical fibroxanthoma.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oculocutaneous Albinism Type 1: Link between Mutations, Tyrosinase Conformational Stability, and Enzymatic Activity
Dolinska MB, et al.
Pigment Cell & Melanoma Research, 30(1), 41-52 (2017)
LEF-1 Regulates Tyrosinase Gene Transcription In Vitro
Wang X, et al.
PLoS ONE, 10(11), e0143142-e0143142 (2015)
Aberrant tyrosinase expression in an atypical fibroxanthoma: A case report.
Weiss A, et al.
Journal of Cutaneous Pathology, 44(5), 467-469 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service