Skip to Content
Merck
All Photos(1)

Key Documents

SAB4502988

Sigma-Aldrich

Anti-TNF Receptor I antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CD120a, TBPI, TNF-RI, p55, p60

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 50 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNFRSF1A(7132)

General description

Tumor necrosis factor receptor 1 (TNFR1) belongs to the TNFR superfamily and is ubiquitously expressed. It comprises an extracellular domain with a cysteine-rich motif, a transmembrane segment, and an intracellular domain with a “death domain” motif. The TNFR1 gene is mapped to human chromosome 12p13.31.

Immunogen

The antiserum was produced against synthesized peptide derived from human TNF Receptor I.

Immunogen Range: 381-430

Application

Anti-TNF Receptor I antibody produced in rabbit has been used in immunostaining at a dilution of 1:200.

Biochem/physiol Actions

Tumor necrosis factor receptor 1 (TNFR1) is crucial for TNF for eliciting proinflammatory functions. Through its dead domain motif, TNFR1 interacts with the TNF receptor-associated death domain protein (TRADD). TNFR1 regulates cytotoxic signaling pathways. Blockage of TNFR1 in mouse model harboring respiratory syncytial virus (RSV) infection results in enhanced bronchoconstriction.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of TNF-TNF Receptor 2 Signal in Regulatory T Cells and Its Therapeutic Implications.
Yang, et al.
Frontiers in Immunology, 9, 784-784 (2019)
Dorothea R Morris et al.
Viruses, 12(10) (2020-10-22)
Respiratory syncytial virus (RSV) is the leading cause of bronchiolitis in infants and young children. Although some clinical studies have speculated that tumor necrosis factor (TNF)-α is a major contributor of RSV-mediated airway disease, experimental evidence remains unclear or conflicting.
The death domain of tumor necrosis factor receptor 1 is necessary but not sufficient for Golgi retention of the receptor and mediates receptor desensitization.
Gaeta, et al.
Laboratory Investigation; a Journal of Technical Methods and Pathology, 80, 1185-1194 (2019)
Ryoko Kikuchi-Koike et al.
BMC cancer, 19(1), 521-521 (2019-05-31)
Patients with lymph node metastasis-negative (pN0) invasive breast cancer have favorable outcomes following initial treatment. However, false negatives which occur during routine histologic examination of lymph nodes are reported to underestimate the clinical stage of disease. To identify a high-risk
Bence Pál Barta et al.
World journal of diabetes, 14(1), 48-61 (2023-01-24)
Cytokines are essential in autoimmune inflammatory processes that accompany type 1 diabetes. Tumor necrosis factor alpha plays a key role among others in modulating enteric neuroinflammation, however, it has a dual role in cell degeneration or survival depending on different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service