Skip to Content
Merck
All Photos(3)

Key Documents

SAB4502493

Sigma-Aldrich

Anti-PTHR1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

PTH1 Receptor, PTH1R, PTHR, Parathyroid hormone 1 receptor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 52 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTH1R(5745)

General description

Anti-PTHR1 antibody detects endogenous levels of total PTHR1 protein.
PTHR1 (parathyroid hormone 1 receptor) gene is mapped to human chromosome 3p21.31. The gene encodes for a precursor (secretin-like class II G protein-coupled receptor) that matures to a receptor containing 593 amino acids. This receptor contains an extracellular loop with a PTH binding site and an intracellular signaling domain. It also includes seven transmembrane and helical segments. The gene is predominantly expressed in osteoblasts and renal tubular cells.

Immunogen

The antiserum was produced against synthesized peptide derived from human PTHR1.

Immunogen Range: 145-194

Biochem/physiol Actions

Heterozygous mutation of PTHR1 (parathyroid hormone 1 receptor) gene is known to cause primary failure of tooth eruption in human, with partial eruption of posterior teeth and reduced alveolar growth process. PTHR1 is associated with bone remodeling (bone formation and resorption, anabolic and catabolic responses respectively) and calcium homeostasis. Blomstrand chondrodysplasia results due to complete loss of PTHR1 function, and is considered lethal.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanghang Liu et al.
eLife, 12 (2023-05-09)
Conditional deletion of the PTH1R in mesenchymal progenitors reduces osteoblast differentiation, enhances marrow adipogenesis, and increases zinc finger protein 467 (Zfp467) expression. In contrast, genetic loss of Zfp467 increased Pth1r expression and shifts mesenchymal progenitor cell fate toward osteogenesis and
A novel homozygous PTH1R variant identified through whole-exome sequencing further expands the clinical spectrum of primary failure of tooth eruption in a consanguineous Saudi family.
Jelani M.
Archives of Oral Biology, 67, 28-33 (2016)
Katja Braun et al.
Physiological reports, 6(17), e13842-e13842 (2018-09-11)
An active lifestyle is generally recommended for hypertensive patients to prevent subsequent end-organ damage. However, experimental data on long-term effects of exercise on hypertension are insufficient and underlying mechanisms are not well understood. This study was aimed to investigate the
VMP1 Establishes ER-Microdomains that Regulate Membrane Contact Sites and Autophagy.
Tabara LC and Escalante R
PLoS ONE, 11(11), 1-20 (2016)
Lei Xiong et al.
EBioMedicine, 9, 45-60 (2016-06-23)
Parathyroid hormone (PTH) plays critical, but distinct, roles in bone remodeling, including bone formation (anabolic response) and resorption (catabolic response). Although its signaling and function have been extensively investigated, it just began to be understood how distinct functions are induced

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service