Skip to Content
Merck
All Photos(3)

Key Documents

SAB4502380

Sigma-Aldrich

Anti-PKC ζ antibody produced in rabbit

affinity isolated antibody

Synonym(s):

KPCZ, PKC-ζ, PKC2, PRKCZ, Protein kinase C ζ type

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 67 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRKCZ(5590)

Related Categories

General description

Protein kinase C (PKC) ϲ is an isoenzyme, which belongs to the atypical class of protein kinase C family. PKCϲ is encoded by the gene mapped to human chromosome 1p36.33-p36.2. PKC ϲ is expressed in human eosinophils. This protein is characterized with a PB1 (Phox and Bem1) domain in the N-terminal, a pseudosubstrate (PS) sequence, a C1 domain (or phorbol esters/diacylglycerol binding domain) of a single Cys-rich zinc-finger motif, and a kinase domain in the C-terminal.

Immunogen

The antiserum was produced against synthesized peptide derived from human PKC zeta.

Immunogen Range: 526-575

Application

Anti-PKC ϲ, C-Terminal antibody produced in rabbit has been used in immunohistochemistry.

Biochem/physiol Actions

Isoenzymes of protein kinase C (PKC) family play a vital role in downstream signaling and cell functions. PKC ϲ regulates adhesion and effector functions of human eosinophils stimulated with platelet-activating factor (PAF) or complement (C) 5a. PKC ϲ and PKC δ might act as a therapeutic target for decreasing eosinophilic inflammation in clinical conditions such as asthma. PKC ϲ is calcium–independent, and is activated by phospholipids including phosphatidic acid (PA), phosphatidylinositol triphosphates and ceramides. Experimental studies state that, PKC ϲ is involved in the mitogen-activated protein kinase (MAPK) cascade, transcriptional factor NF-κB (NF-κB) activation, ribosomal S6-protein kinase signaling, and cell polarity. PKC-ϲ mediates hypoxia-induced Na,K-ATPase endocytosis in the alveolar epithelial cells (AEC).4 PKC-ϲ functions as a potential biomarker for human prostate cancer.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kutibh Chihabi et al.
Behavioral neuroscience, 130(6), 563-571 (2016-03-08)
Protein kinase M zeta (PKM-ζ), a constitutively active N-terminal truncated form of PKC-ζ, has long been implicated in a cellular correlate of learning, long-term potentiation (LTP). Inhibition of PKM-ζ with zeta-inhibitory peptide (ZIP) has been shown in many brain structures
Takaaki Hirai et al.
Journal of biochemistry, 133(1), 1-7 (2003-05-23)
The zeta isotype of protein kinase C (PKCzeta) is a member of the atypical PKC subfamily and has been widely implicated in the regulation of cellular functions. Increasing evidence from studies using in vitro and in vivo systems points to
Andrew C Zelhof et al.
PLoS genetics, 16(6), e1008890-e1008890 (2020-06-25)
The Drosophila apical photoreceptor membrane is defined by the presence of two distinct morphological regions, the microvilli-based rhabdomere and the stalk membrane. The subdivision of the apical membrane contributes to the geometrical positioning and the stereotypical morphology of the rhabdomeres
Masahiko Kato et al.
Immunology, 116(2), 193-202 (2005-09-16)
Protein kinase (PK) C comprises a family of isoenzymes that play key roles in downstream signalling and cell functions. We studied PKC zeta participation in the effector functions of human eosinophils stimulated with platelet-activating factor (PAF) or complement (C) 5a.
Anna G McGrath et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 43(12), 2390-2398 (2018-08-17)
The constitutively active, atypical protein kinase C, protein kinase M-ζ (PKMζ), is exclusively expressed in the brain and its expression increases following exposure to drugs of abuse. However, the limitations of currently available tools have made it difficult to examine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service