Skip to Content
Merck
All Photos(3)

Key Documents

SAB4502167

Sigma-Aldrich

Anti-PECAM-1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CD31, CD31 antigen, EndoCAM, PEC1, PECA1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 82 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PECAM1(5175)

General description

Anti-PECAM-1 Antibody detects endogenous levels of total PECAM-1 protein.
The platelet and endothelial cell adhesion molecule 1 (PECAM1) gene, spanning 75-kb, is mapped on the long arm of human chromosome 17. This gene codes for 130-kD protein, which belongs to the immunoglobulin (Ig) superfamily. It is a vital component of the endothelial cell intercellular junction. PECAM1 is expressed on the surface of circulating platelets, monocytes, neutrophils and selected T-cell subsets.

Immunogen

The antiserum was produced against synthesized peptide derived from human PECAM-1.

Immunogen Range: 686-735

Application

Anti-PECAM-1, C-Terminal antibody produced in rabbit has been used in immunohistochemistry.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Platelet and endothelial cell adhesion molecule 1 (PECAM1) also called as cluster of differentiation 31 (CD31), plays a crucial role in the adhesion cascade leading to leukocyte extravasation during the inflammatory process. PECAM1 is essential for leukocyte transmigration via intercellular junctions of vascular endothelial cells. Genetic polymorphism of PECAM1 is associated with atherosclerotic events. PECAM1, is a membrane glycoprotein that facilitates both homophilic and heterophilic adhesion. PECAM1 is also implicated in angiogenesis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charan Thej et al.
Stem cell research & therapy, 12(1), 279-279 (2021-05-12)
We have previously demonstrated that a pooled population of bone marrow-derived, allogeneic mesenchymal stromal cells (BMMSC), Stempeucel®-1, produced under good manufacturing practices (GMP) conditions, showed clinical efficacy and safety in patients suffering from critical limb ischemia (CLI) due to Buerger's
Guorong Li et al.
Investigative ophthalmology & visual science, 61(14), 12-12 (2020-12-15)
Tyrosine kinase with immunoglobulin-like and EGF-like domains 2 (Tie2) activation in Schlemm's canal (SC) endothelium is required for the maintenance of IOP, making the angiopoietin/Tie2 pathway a target for new and potentially disease modifying glaucoma therapies. The goal of the
Omer Nuri Pamuk et al.
Clinical rheumatology, 33(12), 1737-1743 (2014-09-10)
Genetic polymorphisms of platelet endothelial cell adhesion molecule-1 (PECAM-1) were found to play roles in atherosclerotic events. We determined PECAM-1 polymorphisms, soluble PECAM-1, and CD40L levels in rheumatoid arthritis (RA) and systemic lupus erythematosus (SLE) and evaluated their associations with
Ge Li et al.
Cell death & disease, 8(5), e2811-e2811 (2017-05-26)
Programmed cell death 5 (PDCD5) is an apoptosis promoter molecule that displays multiple biological activities. However, the function of PDCD5 in vivo has not yet been investigated. Here, we generated a Pdcd5 knockout mouse model to study the physiological role
Xiao-Jun Zheng et al.
The Journal of pathology, 248(4), 438-451 (2019-03-23)
The mineralocorticoid receptor (MR) plays important roles in cardiovascular pathogenesis. The function of MR in angiogenesis is still controversial. This study aimed to explore the role of endothelial MR in angiogenesis and to delineate the underlying mechanism. Endothelial-hematopoietic MR knockout

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service