Skip to Content
Merck
All Photos(2)

Documents

SAB4501137

Sigma-Aldrich

Anti-Glucagon antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Glicentin, Glicentin-related polypeptide, Glucagon-like peptide 1(7-37), Glucagon-like peptide 1(GLP-1), Oxyntomodulin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 20 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GCG(2641)

General description

Anti-Glucagon antibody detects endogenous levels of total Glucagon protein.
Glucagon-like peptide-1 (GLP-1) is a gastrointestinal hormone. It is a 30-amino acid peptide, that is generated by the intestinal epithelial endocrine L-cells. it is mapped to human chromosome 2q24.

Immunogen

The antiserum was produced against synthesized peptide derived from human Glucagon.

Immunogen Range: 61-110

Application

Anti-glucagon antibody has been used in immunostaining and immunohistochemistry.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Glucagon-like peptide-1 (GLP-1) increases glucose-dependent insulin secretion and terminates the liberation of glucagon. It acts as a physiological regulator of appetite and food intake. GLP-1 also helps to prevent the gastrointestinal movement and secretion.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Physiology of Glucagon-like Peptide 1
Holst JJ
American Physiological Society, 87(4), 1409-1439 (2007)
Expression and regulation of chemokines in murine and human type 1 diabetes
Sarkar SA, et al.
Diabetes, 61(2), 436-446 (2012)
Reg3? Overexpression Protects Pancreatic Beta-Cells From Cytokine-Induced Damage and Improves Islet Transplant Outcome
Ding Y, et al.
Molecular Medicine, 20(1), 548-548 (2014)
Glucagon-like Peptide-1 Analogues Inhibit Proliferation and Increase Apoptosis of Human Prostate Cancer Cells in vitro
Li XN, et al.
Experimental and Clinical Endocrinology & Diabetes, 125(02), 91-97 (2017)
Maria Eriksson et al.
Molecular pharmaceutics, 18(12), 4428-4436 (2021-10-16)
Type 1 diabetes develops in childhood and adolescence, with peak incidence in the early teenage years. There is an urgent need for an accurate method to detect insulin-producing β-cells in patients that is not affected by alterations in β-cell function.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service