Skip to Content
Merck
All Photos(3)

Documents

SAB4500435

Sigma-Aldrich

Anti-Claudin 3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CPE-R 2, CPE-receptor 2, Claudin-3, Clostridium perfringens enterotoxin receptor 2, Ventral prostate.1 protein homolog

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 23 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLDN3(1365)

Related Categories

General description

Anti-Claudin 3 Antibody detects endogenous levels of total Claudin 3 protein.
Claudin-3 (CLDN3) is a tetraspanin transmembrane protein, that is mapped to human chromosome 7q11.23.

Immunogen

The antiserum was produced against synthesized peptide derived from human Claudin 3.

Immunogen Range: 171-220

Application

Anti-Claudin 3, C-Terminal antibody has been used in immunofluorescence and western blotting.

Biochem/physiol Actions

In submandibular gland, claudin-3 (CLDN3) controls paracellular permeability with the help of TNF-α(tumor necrosis factor α). High level expression of CLDN3 improves the malignant ability of colorectal cancer cells. It participates in the organization and activities of tight junctions (TJs).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudin-3 is required for modulation of paracellular permeability by TNF-? through ERK1/2/slug signaling axis in submandibular gland
Mei M, et al.
Cellular Signalling, 27(10), 1915-1927 (2015)
Generation of specific monoclonal antibodies against the extracellular loops of human claudin-3 by immunizing mice with target-expressing cells
Ando H, et al.
Bioscience, Biotechnology, and Biochemistry, 79(8), 1272-1279 (2015)
Evidence for a role of claudin 2 as a proximal tubular stress responsive paracellular water channel
Wilmes A, et al.
Toxicology and Applied Pharmacology, 279(2), 163-172 (2014)
Claudin-3 overexpression increases the malignant potential of colorectal cancer cells: roles of ERK1/2 and PI3K-Akt as modulators of EGFR signaling
de Souza WF, et al.
PLoS ONE (2013)
Katharina M C Klee et al.
Human genetics, 139(10), 1247-1259 (2020-04-20)
Congenital diarrheal disorders (CDD) comprise > 50 monogenic entities featuring chronic diarrhea of early-onset, including defects in nutrient and electrolyte absorption, enterocyte polarization, enteroendocrine cell differentiation, and epithelial integrity. Diarrhea is also a predominant symptom in many immunodeficiencies, congenital disorders of glycosylation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service