Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500429

Sigma-Aldrich

Anti-ADORA2B antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ADORA2B, Adenosine receptor A2b

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 36 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADORA2B(136)

General description

Anti-ADORA2B Antibody detects endogenous levels of total ADORA2B protein.
ADORA2B (adenosine A2b receptor) is a G-protein coupled receptor. It is the most adenosine insensitive adenosine receptor. This gene is located on human chromosome 17p12-p11.2. Anti-ADORA2B Antibody detects endogenous levels of total ADORA2B protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human ADORA2B.

Immunogen Range: 231-280

Biochem/physiol Actions

ADORA2B (adenosine A2b receptor) acts as a therapeutic target for cancer treatment. The tumor cells can use A2b receptor to induce tumor progression. ADORA2B controls immune cell activity. It plays a major role in the transcriptional induction and protein stability of protein period 2 (Per2).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CD73-Dependent Generation of Adenosine and Endothelial Adora2b Signaling Attenuate Diabetic Nephropathy.
Eunyoung T, et al.
Journal of the American Society of Nephrology, 25 (3), 547-563 (2014)
Adora2b-elicited Per2 stabilization promotes a HIF-dependent metabolic switch crucial for myocardial adaptation to ischemia.
Eckle T, et al.
Nature Medicine, 18(5), 774-774 (2012)
Pharmacogenetics of asthma.
Fenech A and Hall IP
British Journal of Clinical Pharmacology, 53(1), 3-15 (2002)
Role of adenosine A2b receptor overexpression in tumor progression.
Sepulveda C, et al.
Life Sciences, 166, 92-99 (2016)
The A2b adenosine receptor antagonist PSB-603 promotes oxidative phosphorylation and ROS production in colorectal cancer cells via adenosine receptor-independent mechanism.
M?lck C, et al.
Cancer Letters, 383(1), 135-143 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service