Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500352

Sigma-Aldrich

Anti-Histone H3.1 Antibody

rabbit polyclonal

Synonym(s):

H3/a, H3/c, H3/d, H3/f, H3/h

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

Anti-Histone H3 antibody produced in rabbit, affinity isolated antibody

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 15 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HIST1H3A(8350)

General description

Anti-Histone H3 Antibody detects endogenous levels of total Histone H3 protein.
Mammals contain three main classes of histone H3 variants: the replicative histones (H3.1 and H3.2), the replacement histone (H3.3) and the centromeric histone (Cenp-A). Histone H3 is located on human chromosome 6p22.

Immunogen

The antiserum was produced against synthesized peptide derived from human Histone H3.

Immunogen Range: 1-50

Application

Anti-Histone H3, N-Terminal antibody has been used in western blotting and enzyme-linked immunosorbent assay (ELISA).

Biochem/physiol Actions

Histone proteins are basic building blocks of chromatin. Histone H3 K27M mutations in adult cerebellar high-grade gliomas. It controls protein-protein interactions to induce binding of trans-acting factors that “drive” chromatin condensation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Histone Demethylation and Toll?like Receptor 8?Dependent Cross?Talk in Monocytes Promotes Transdifferentiation of Fibroblasts in Systemic Sclerosis Via Fra?2.
Ciechomska M, et al.
Arthritis and Rheumatism, 68(6), 1493-1504 (2016)
Dihydromyricetin protects against liver ischemia/reperfusion induced apoptosis via activation of FOXO3a-mediated autophagy.
Chen Y, et al.
Oncotarget, 7(47), 76508-76508 (2016)
Parissa C Monem et al.
PLoS genetics, 19(1), e1010577-e1010577 (2023-01-11)
As ribosomes translate the genetic code, they can encounter a variety of obstacles that hinder their progress. If ribosomes stall for prolonged times, cells suffer due to the loss of translating ribosomes and the accumulation of aberrant protein products. Thus
Omar Ramírez-Nuñez et al.
Journal of neurochemistry, 158(2), 482-499 (2021-04-28)
Nucleocytosolic transport, a membrane process, is impaired in motor neurons in amyotrophic lateral sclerosis (ALS). This study analyzes the nuclear lipidome in motor neurons in ALS and examines molecular pathways linked to the major lipid alterations. Nuclei were obtained from
Yongbiao Chen et al.
Oncotarget, 7(47), 76508-76522 (2016-10-30)
Liver ischemia and reperfusion (I/R) injury is characterized by defective liver autophagy accompanied by alterations to the endogenous defense system. Dihydromyricetin (DHM) is a natural flavonoid that demonstrates a wide range of physiological functions, and has been implicated as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service