Skip to Content
Merck
All Photos(1)

Documents

SAB4300693

Sigma-Aldrich

Anti-CDH1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-CD324, Anti-CDHE, Anti-E-Cadherin, Anti-UVO

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~135 kDa

species reactivity

mouse, rat, human

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(E-W-G-N-R)

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDH1(999)

General description

CDH1 (cadherin 1) codes for a transmembrane glycoprotein E-cadherin that is located on human chromosome 16q22.1. It is a tumor suppressor gene, which is also known as Cdh1p/Hct1p, Fizzy-related, Ste9, or Srw1.
The antibody detects endogenous level of total E-Cadherin protein.

Immunogen

Peptide sequence around aa. 864-868 (E-W-G-N-R), according to the protein NP_004351.1

Biochem/physiol Actions

CDH1 (cadherin 1) helps to conserve the homophilic cell-cell adhesion in epithelial tissues. Inadequate expression of CDH1 results in colon cancer and renal cell carcinoma (RCC).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Cadherins are calcium-dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells; cadherins may thus contribute to the sorting of heterogeneous cell types. CDH1 is involved in mechanisms regulating cell-cell adhesions, mobility and proliferation of epithelial cells. Has a potent invasive suppressor role. It is a ligand for integrin alpha-E/beta-7. E-Cad/CTF2 promotes non-amyloidogenic degradation of Abeta precursors. Has a strong inhibitory effect on APP C99 and C83 production.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CDH1 promoter hypermethylation and E-cadherin protein expression in infiltrating breast cancer
Caldeira JR, et al.
BMC Cancer, 6, 48-48 (2006)
Identification of downstream metastasis-associated target genes regulated by LSD1 in colon cancer cells
Chen J, et al.
Oncotarget, 8(12), 19609-19630 (2017)
Reduced E-cadherin facilitates renal cell carcinoma progression by WNT/?-catenin signaling activation
Zhang X, et al.
Oncotarget, 8(12), 19566-19576 (2017)
Mitotic regulation of the APC activator proteins CDC20 and CDH1
Kramer ER, et al.
Molecular Biology of the Cell, 11(5), 1555-1569 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service