Skip to Content
Merck
All Photos(2)

Key Documents

SAB4300337

Sigma-Aldrich

Anti-PTEN (Ab-370) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-10q23del antibody produced in rabbit, Anti-BZS antibody produced in rabbit, Anti-MGC11227 antibody produced in rabbit, Anti-MHAM antibody produced in rabbit, Anti-phosphatase and tensin homolog antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~54 kDa

species reactivity

mouse, human, rat

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(D-V-S-D-N)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTEN(5728)

Immunogen

Peptide sequence around aa. 368-372 (D-V-S-D-N), according to the protein PTEN.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Tumor suppressor. Acts as a dual-specificity protein phosphatase, dephosphorylating tyrosine-, serine- and threonine-phosphorylated proteins. Also acts as a lipid phosphatase, removing the phosphate in the D3 position of the inositol ring from phosphatidylinositol 3,4,5-trisphosphate, phosphatidylinositol 3,4-diphosphate, phosphatidylinositol 3-phosphate and inositol 1,3,4,5-tetrakisphosphate with order of substrate preference in vitro PtdIns(3,4,5)P3 > PtdIns(3,4)P2 > PtdIns3P > Ins(1,3,4,5)P4. The lipid phosphatase activity is critical for its tumor suppressor function. Antagonizes the PI3K-AKT/PKB signaling pathway by dephosphorylating phosphoinositides and thereby modulating cell cycle progression and cell survival. The unphosphorylated form cooperates with AIP1 to suppress AKT1 activation. Dephosphorylates tyrosine-phosphorylated focal adhesion kinase and inhibits cell migration and integrin-mediated cell spreading and focal adhesion formation. May be a negative regulator of insulin signaling and glucose metabolism in adipose tissue.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hang-Zhi Gu et al.
Oncology letters, 13(5), 3032-3038 (2017-05-20)
Endometrial carcinoma (EC) is one of the most common female malignancies, and there is an urgent requirement to explore new therapeutic strategies. In the present study, Ishikawa H cells were treated with Momordica charantia protein (MCP30). The cell morphology, growth
Jianxin Yan et al.
Stem cell research & therapy, 12(1), 220-220 (2021-04-02)
Mesenchymal stem cells (MSCs) exert positive effects in chronic wounds. However, critical parameters, such as the most effective administration routes, remain unclear. Accordingly, the purpose of this study was to compare the effects of topical and systemic transplantation MSCs on
Lili Liu et al.
Bioscience reports, 40(5) (2020-05-19)
The present study aims to reveal the molecular mechanism of peroxisome proliferator-activated receptor γ (PPARγ) on sepsis-induced acute lung injury (ALI). To do that, the rat injury model was established using cecal ligation and perforation (CLP) method, followed by different
Shupeng Wu et al.
Molecular medicine reports, 17(6), 7966-7972 (2018-04-06)
There are several reports in the literature regarding microRNA (miR)‑130b. It has been reported that miR‑130b is involved in several diseases. The present study aimed to understand the association between the levels of miR‑130b and lupus nephritis in patients. A
Ming Tu et al.
Molecular medicine reports, 15(4), 1713-1721 (2017-03-06)
Cluster of differentiation 164 (CD164), a sialomucin, has been demonstrated to be involved in the regulation of proliferation, apoptosis, adhesion and differentiation in multiple cancers. CD164 is regarded to be a potential promotor of tumor growth. However, the involvement of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service