Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300251

Sigma-Aldrich

Anti-phospho-SMAD2 (pSer467) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-JV18 antibody produced in rabbit, Anti-JV18-1 antibody produced in rabbit, Anti-MADH2 antibody produced in rabbit, Anti-MADR2 antibody produced in rabbit, Anti-SMAD family member 2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~60 kDa

species reactivity

rat, human, mouse

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(C-S-S-M-SP)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer467)

Gene Information

human ... SMAD2(4087)

Immunogen

Peptide sequence around phosphorylation site of serine 467 (C-S-S-M-S(p)), according to the protein SMAD2.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Transcriptional modulator activated by TGF-beta and activin type 1 receptor kinase. SMAD2 is a receptor-regulated SMAD (R-SMAD). May act as a tumor suppressor in colorectal carcinoma.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuejiao Wei et al.
Molecular medicine reports, 17(1), 495-501 (2017-11-09)
The aim of the present study was to analyze the effect of diltiazem on myocardial fibrosis and remodeling of connexin43 (Cx43) in myocardial ischemic rats and mechanisms underlying these processes. A total of 36 Sprague‑Dawley rats were randomly allocated into
Xiuli Zhang et al.
Kidney & blood pressure research, 43(2), 500-512 (2018-04-09)
Evidence from our and other groups has demonstrated that zinc transporter 7 in SLC30 family (ZnT7) inhibited epithelial-to-mesenchymal transition (EMT) and apoptosis in rat peritoneal mesothelial cells (RPMCs) under high glucose (HG) concentration. In the present study, we investigated the
He-Song Jiang et al.
International journal of molecular medicine, 36(3), 801-807 (2015-07-17)
The transformation of tunica albuginea-derived fibroblasts (TAFs) into myofibroblasts plays an important role in the pathological progress of Peyronie's disease (PD). However, no treatment which addresses this transformation is currently available. Estrogen has been shown to inhibit the progression of
Milena Paw et al.
International journal of molecular sciences, 19(9) (2018-08-31)
The activation of human bronchial fibroblasts by transforming growth factor-β₁ (TGF-β₁) leads to the formation of highly contractile myofibroblasts in the process of the fibroblast⁻myofibroblast transition (FMT). This process is crucial for subepithelial fibrosis and bronchial wall remodeling in asthma.
Lingfang Zeng et al.
The Journal of biological chemistry, 288(44), 31853-31866 (2013-09-21)
Histone deacetylase 3 (HDAC3) plays a critical role in the maintenance of endothelial integrity and other physiological processes. In this study, we demonstrated that HDAC3 undergoes unconventional splicing during stem cell differentiation. Four different splicing variants have been identified, designated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service