Skip to Content
Merck
All Photos(2)

Documents

SAB4300032

Sigma-Aldrich

Anti-phospho-STAT1 (pTyr701) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp686B04100 antibody produced in rabbit, Anti-ISGF-3 antibody produced in rabbit, Anti-STAT91 antibody produced in rabbit, Anti-signal transducer and activator of transcription 1, 91kDa antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~91 kDa

species reactivity

human, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(T-G-YP-I-K)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr701)

Gene Information

human ... STAT1(6772)

General description

Signal transducer and activator of transcription 1 (STAT1) gene is mapped to human chromosome 2q32.2. It is part of the STAT family of proteins. STAT1 exists in two isoforms, STAT1α and STAT1β with molecular weight of 91kDa and 84kDa, respectively. The protein shows ubiquitous expression.

Immunogen

Peptide sequence around phosphorylation site of tyrosine 701 (T-G-Y(p)-I-K), according to the protein STAT1.

Biochem/physiol Actions

Signal transducer and activator of transcription 1 (STAT1) plays a vital role in interferon (IFN) signaling. Phospho-STAT1 regulates pro- and anti-apoptotic genes following several stress-induced responses. It interacts with p53 and promotes p53-mediated transcriptional gene activity and apoptosis. STAT1 is also an essential factor of stress-induced and DNA damage-induced checkpoint pathways and thus, acts as a tumor suppressor.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Epigallocatechin-3-gallate inhibits STAT-1 activation and protects cardiac myocytes from ischemia/reperfusion-induced apoptosis.
Faseb Journal (2004)
STAT-1: a novel regulator of apoptosis.
Stephanou A and Latchman DS
International Journal of Experimental Pathology (2003)
cDNA cloning, expression and chromosome mapping of the human STAT4 gene: both STAT4 and STAT1 genes are mapped to 2q32.2-->q32.3.
Yamamoto K
Cytogenetics and Cell Genetics (1997)
Assignment1 of STAT1 to human chromosome 2q32 by FISH and radiation hybrids.
Haddad B
Cytogenetics and Cell Genetics (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service