Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200543

Sigma-Aldrich

Anti-Calbindin-D-28K antibody, Mouse monoclonal

enhanced validation

clone CB-955, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-(28kD), Monoclonal Anti-D-28K, Monoclonal Anti-D-dependent calcium-binding protein, Monoclonal Anti-RTVL-H protein, Monoclonal Anti-avian-type, Monoclonal Anti-calbindin 1, Monoclonal Anti-vitamin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CB-955, monoclonal

form

buffered aqueous solution

mol wt

antigen ~28 kDa

species reactivity

mouse, human, rat

enhanced validation

independent
Learn more about Antibody Enhanced Validation

concentration

~1.0 mg/mL

technique(s)

immunohistochemistry: 10-20 μg/mL using formalin-fixed, paraffin embedded rat cerebellum.
western blot: 2-4 μg/mL using MDBK extract.

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CALB1(793)

General description

Anti-Calbindin-D-28K antibody, Mouse monoclonal (mouse IgG1 isotype) is derived from the hybridoma CB-955 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a purified bovine kidney calbindin-D-28K.
Calbindin-D-28K (CALB1) is encoded by the gene mapped to human chromosome 8q21.3-q22.1. CALB1 belongs to the EF hand superfamily of Ca2+ binding proteins (CBPs). The encoded protein contains EF hand motif involved in the specific and reversible binding of Ca2+. It is highly expressed in specific regions of central nervous system such as cerebellar Purkinje cells and hippocampal granule cells.

Immunogen

purified bovine kidney calbindin-D-28K. The isotype is determined by ELISA using Mouse Monoclonal Antibody Isotyping Reagents (Sigma ISO-2). The antibody is purified from culture supernatant of hybridoma cells grown in a bioreactor.

Application

  • Anti-Calbindin-D-28K antibody, Mouse monoclonal has been used in:
  • western blotting
  • enzyme-linked immunosorbent assay (ELISA){31
  • immunohistochemistry
  • immunohistochemical procedure
  • indirect immunofluorescence

Biochem/physiol Actions

Calbindin D-28k might play a vital role as an intracellular Ca2+ buffer. It has an ability to protect nerve cells from calcium-mediated neurotoxicity. The encoded protein functions as a potent neuroanatomical marker which can be used to selectively visualize certain neurons and pathways in the central nervous system and peripheral nervous system. Mutations in the gene has been observed in patients with Parkinson′s disease (PD).
Calbindin-D (28K) functions as a mobile or partly immobilized Ca2+ -buffer with medium kinetics and affinity buffer function. Second, it functions in buffered Ca2+- diffusion-transport function. Finally, it interacts with target proteins in its apo-and Ca2+-loaded form-sensor- like function. Over-expression of this protein stimulates neurite outgrowth in dopaminergic neuronal cells.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A comparative study of three interneuron types in the rat spinal cord
Chen S, et al.
PLoS ONE, 11(9), e0162969-e0162969 (2016)
Ai Takeshita et al.
Scientific reports, 8(1), 6917-6917 (2018-05-04)
Fibroblast growth factor 23 (FGF23) plays critical roles in phosphate handling and vitamin D metabolism in the kidney. However, the effector cells of FGF23 in the kidney remain unclear. αKlotho, a putative enzyme possessing β-glucuronidase activity and also a permissive
The human calbindin D28k (CALB1) and calretinin (CALB2) genes are located at 8q21.3----q22.1 and 16q22----q23, respectively, suggesting a common duplication with the carbonic anhydrase isozyme loci.
Parmentier M
Cytogenetics and Cell Genetics, 57, 41-43 (1991)
Binding Kinetics of Calbindin-D28k Determined by Flash Photolysis of Caged Ca2+
Nagerl UV
Biophysical Journal, 79, 3009-3018 (2000)
Overexpression of calbindin-D28K induces neurite outgrowth in dopaminergic neuronal cells via activation of p38 MAPK
Choi WS, et al.
Biochemical and biophysical research communications, 287(3), 656-661 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service