Skip to Content
Merck
All Photos(3)

Documents

SAB3500979

Sigma-Aldrich

Anti-ACE2 antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

ELISA: suitable
immunoblotting: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... ACE2(59272)

Immunogen

Antibody was raised against a synthetic peptide corresponding to amino acids near the center of human ACE2.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500979.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael A Crackower et al.
Nature, 417(6891), 822-828 (2002-06-21)
Cardiovascular diseases are predicted to be the most common cause of death worldwide by 2020. Here we show that angiotensin-converting enzyme 2 (ace2) maps to a defined quantitative trait locus (QTL) on the X chromosome in three different rat models
S R Tipnis et al.
The Journal of biological chemistry, 275(43), 33238-33243 (2000-08-05)
A novel human zinc metalloprotease that has considerable homology to human angiotensin-converting enzyme (ACE) (40% identity and 61% similarity) has been identified. This metalloprotease (angiotensin-converting enzyme homolog (ACEH)) contains a single HEXXH zinc-binding domain and conserves other critical residues typical
M Donoghue et al.
Circulation research, 87(5), E1-E9 (2000-09-02)
ACE2, the first known human homologue of angiotensin-converting enzyme (ACE), was identified from 5' sequencing of a human heart failure ventricle cDNA library. ACE2 has an apparent signal peptide, a single metalloprotease active site, and a transmembrane domain. The metalloprotease
Wenhui Li et al.
Nature, 426(6965), 450-454 (2003-12-04)
Spike (S) proteins of coronaviruses, including the coronavirus that causes severe acute respiratory syndrome (SARS), associate with cellular receptors to mediate infection of their target cells. Here we identify a metallopeptidase, angiotensin-converting enzyme 2 (ACE2), isolated from SARS coronavirus (SARS-CoV)-permissive
Zhen Zou et al.
Nature communications, 5, 3594-3594 (2014-05-08)
The potential for avian influenza H5N1 outbreaks has increased in recent years. Thus, it is paramount to develop novel strategies to alleviate death rates. Here we show that avian influenza A H5N1-infected patients exhibit markedly increased serum levels of angiotensin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service