Skip to Content
Merck
All Photos(3)

Key Documents

SAB3500384

Sigma-Aldrich

Anti-MANF antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ARMET, Anti-ARP, Anti-Arginine-rich mutatedin early stage tumor, Anti-Mesencephalic astrocyte-derived neurotrophic factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 20 kDa

species reactivity

rat, human, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ARMET(7873)

Immunogen

MANF antibody was raised against a 12 amino acid peptide from near the carboxy terminus of human MANF.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500384.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiamin Yan et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 36(6), e22349-e22349 (2022-05-15)
Excessive lipid accumulation, inflammation, and fibrosis in the liver are the major characteristics of non-alcoholic steatohepatitis (NASH). Mesencephalic astrocyte-derived neurotrophic factor (MANF) plays an important role in metabolic homeostasis, raising the possibility that it is involved in NASH. Here, we
Tong Wu et al.
The Journal of experimental medicine, 218(6) (2021-04-16)
Activating beige adipocytes in white adipose tissue (WAT) to increase energy expenditure is a promising strategy to combat obesity. We identified that mesencephalic astrocyte-derived neurotrophic factor (Manf) is a feeding-induced hepatokine. Liver-specific Manf overexpression protected mice against high-fat diet-induced obesity
Michael Peled et al.
PloS one, 16(4), e0250178-e0250178 (2021-04-24)
The most successful immunotherapeutic agents are blocking antibodies to either programmed cell death-1 (PD-1), an inhibitory receptor expressed on T lymphocytes, or to its ligand, programmed cell death-ligand 1 (PD-L1). Nevertheless, many patients do not respond, and additional approaches, specifically
Maricris Bautista et al.
eNeuro, 10(1) (2022-12-17)
Injuries to peripheral nerves are frequent, yet no drug therapies are available for effective nerve repair. The slow growth rate of axons and inadequate access to growth factors challenge natural repair of nerves. A better understanding of the molecules that
Joana Neves et al.
Experimental gerontology, 134, 110893-110893 (2020-03-03)
Ageing is a major risk factor for vision loss, and inflammation is an important contributor to retinal disease in the elderly. Regenerative medicine based on cell replacement strategies has emerged in recent years as a promising approach to restore vision.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service