Skip to Content
Merck
All Photos(5)

Key Documents

SAB3500199

Sigma-Aldrich

Anti-Clusterin antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ApoJ, Anti-Apolipoprotein J, Anti-CLI, Anti-Complement lysis inhibitor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLU(1191)

General description

The Clusterin (CLU) gene is mapped to human chromosome 8p21.1. The encoded protein is a chaperone and is conserved along with evolution. It is present in human tissues and fluids.

Immunogen

Clusterin antibody was raised recombinant human Clusterin isoform 1.

Application

Anti-Clusterin antibody produced in rabbit has been used in immunohistochemical (IHC) staining (1:1,000 & 1:100).

Biochem/physiol Actions

Clusterin (CLU) is a multifunctional plasma protein and a fluid-phase complement inhibitor. It mediates the complement terminal pathway. Clusterin is an ATP-independent small heat shock protein-like chaperone. It functions as a redox sensor and a lipid transporter. Clusterin is associated with many age-related diseases such as neurodegeneration and metabolic syndrome. It is involved in cancer progression, promotion, metastasis, and chemoresistance. Increased serum CLU levels are observed in type 2 diabetes (T2D). It is known to suppress proteotoxicity. Clusterin might show anti or proapoptotic activities. This protein is associated with different functions of the brain. A single nucleotide polymorphism (SNP) in the CLU gene might cause the development of late-onset Alzheimer′s disease (LOAD).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Prakash Priyadarshi Praharaj et al.
Biochimica et biophysica acta. Reviews on cancer, 1875(2), 188500-188500 (2021-01-02)
Clusterin (CLU) is an evolutionary conserved molecular chaperone present in different human tissues and fluids and established to be a significant cancer regulator. It controls several cancer-associated cellular events, including cancer cell proliferation, stemness, survival, metastasis, epithelial-mesenchymal transition, therapy resistance
Katina M Athanas et al.
Schizophrenia research, 169(1-3), 381-385 (2015-10-21)
The expression of the gene that encodes clusterin, a glycoprotein that has been implicated in the regulation of many cellular processes, has previously been found in gene expression profiling studies to be among the most significantly differentially expressed genes in
Sarah M Carpanini et al.
Genes, 12(3) (2021-04-04)
Late-onset Alzheimer's disease (LOAD), the most common cause of dementia, and a huge global health challenge, is a neurodegenerative disease of uncertain aetiology. To deliver effective diagnostics and therapeutics, understanding the molecular basis of the disease is essential. Contemporary large
Ahmed A Moustafa et al.
Reviews in the neurosciences, 29(1), 21-38 (2017-09-28)
In this review, we discuss the genetic etiologies of Alzheimer's disease (AD). Furthermore, we review genetic links to protein signaling pathways as novel pharmacological targets to treat AD. Moreover, we also discuss the clumps of AD-m ediated genes according to
Vladimir J Balcar et al.
Neurochemical research, 46(2), 411-422 (2020-11-19)
Clusterin (CLU; also known as apolipoprotein J, ApoJ) is a protein of inconstant structure known to be involved in diverse processes inside and outside of brain cells. CLU can act as a protein chaperon or protein solubilizer, lipid transporter as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service