Skip to Content
Merck
All Photos(2)

Key Documents

SAB2501088

Sigma-Aldrich

Anti-USH1C/Harmonin antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-AIE-75, Anti-DFNB18, Anti-Harmonin, Anti-PDZ-45, Anti-PDZ-73

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... USH1C(10083)

General description

The Usher syndrome type 1C (USH1C) gene encodes for harmonin, a six PSD95/Dlg/Z-1 homology (PDZ) domain-containing protein. The USH1C gene is mapped on the human chromosome at 11p15.1.

Immunogen

Peptide with sequence DRKVAREFRHKVD-C from the N Terminus of the protein sequence according to NP_005700.2; NP_710142.1.

Biochem/physiol Actions

Harmonin acts as a scaffolding protein and interacts via its six PSD95/Dlg/Z-1 homology (PDZ)-domain with myosin VIIa, scaffold protein containing ankyrin repeats, and SAM domain (SANS), and cadherin 23. It plays a role in normal mechanosensory function in cochlear hair cells. Mutations in the USH1C gene are associated with non-syndromic recessive deafness, retinitis pigmentosa, and Usher syndrome type 1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao Mei Ouyang et al.
Human genetics, 111(1), 26-30 (2002-07-24)
We have recently shown that USH1C underlies Usher syndrome type 1c (USH1C), an USH1 subtype characterized by profound deafness, retinitis pigmentosa, and vestibular dysfunction. USH1C encodes a PDZ-domain-containing protein, harmonin. Eight different Ush1c transcripts were identified in the mouse inner
USH1A: chronicle of a slow death.
Gerber, et al.
American Journal of Human Genetics, 78, 357-359 (2020)
Samer Khateb et al.
PloS one, 7(12), e51566-e51566 (2012-12-20)
We used a combined approach of homozygosity mapping and whole exome sequencing (WES) to search for the genetic cause of autosomal recessive retinitis pigmentosa (arRP) in families of Yemenite Jewish origin. Homozygosity mapping of two arRP Yemenite Jewish families revealed
Jan Reiners et al.
Molecular vision, 11, 347-355 (2005-06-02)
The human Usher syndrome (USH) is the most common form of deaf-blindness. Usher type I (USH1), the most severe form, is characterized by profound congenital deafness, constant vestibular dysfunction and prepubertal onset of retinitis pigmentosa. Five corresponding genes of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service