Skip to Content
Merck
All Photos(1)

Key Documents

SAB2500452

Sigma-Aldrich

Anti-GATA1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ERYF1, Anti-GF1, Anti-HGNC:4170, Anti-NFE1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, canine, mouse, rat

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GATA1(2623)

General description

GATA binding protein 1 (GATA1) is a key hematopoietic transcription factor. It is part of the GATA transcription factor family and the gene encoding it is localized on human chromosome Xp11.23.

Immunogen

Peptide with sequence C-DAEAYRHSPVFQ from the internal region of the protein sequence according to NP_002040.1.

Biochem/physiol Actions

GATA binding protein 1 (GATA1) plays a crucial role in erythroid and megakaryocyte differentiation and proliferation. It modulates the expression of genes and acts as an E-cadherin repressor. Mutations in the GATA1 gene have been linked to transient abnormal myelopoiesis (TAM) and myeloid leukemia of DS (ML-DS). The protein has anti-apoptotic functions in breast cancer.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

GATA1 induces epithelial-mesenchymal transition in breast cancer cells through PAK5 oncogenic signaling.
Li Y
Oncotarget (2015)
GATA1 mutations in a cohort of Malaysian children with Down syndrome-associated myeloid disorder.
Lum SH
Singapore Medical Journal, 57(6), 320-324 (2016)
Leif S Ludwig et al.
Cell reports, 27(11), 3228-3240 (2019-06-13)
Human erythropoiesis serves as a paradigm of physiologic cellular differentiation. This process is also of considerable interest for better understanding anemias and identifying new therapies. Here, we apply deep transcriptomic and accessible chromatin profiling to characterize a faithful ex vivo human erythroid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service