Skip to Content
Merck
All Photos(2)

Documents

SAB2500438

Sigma-Aldrich

Anti-G6PD (AA 308-320) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-G6PD1, Anti-Glucose-6-phosphate dehydrogenase

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

canine, rat, mouse, human

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... G6PD(2539)

Related Categories

General description

The gene encoding G6PD (glucose-6-phosphate dehydrogenase) is mapped to human chromosome Xq28 and spans 16.2 kb. One G6PD monomer is composed of 515 amino acids with a predicted molecular weight of 59,256 Da. The active enzyme exists as a dimer and contains an NADP molecule tightly bound to it.

Immunogen

Peptide with sequence C-STNSDDVRDEKVK from the internal region of the protein sequence according to NP_000393.4 ; NP_001035810.1.

Biochem/physiol Actions

G6PD (glucose-6-phosphate dehydrogenase) enzyme catalyzes the oxidation of glucose-6-phosphate to 6-phosphogluconolactone, reducing NADP to NADPH. It therefore, catalyzes the first step of hexose monophosphate pathway (HMP). As it oxidizes glucose-6-phosphate, it confers protection against oxidative damage in erythrocytes. Its deficiency is highly heterogeneous, with around 190 variants being reported. The deficiency of G6PD was discovered while studying hemolytic anemia. G6PD also results in hereditary nonspherocytic hemolytic anemia.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service