Skip to Content
Merck
All Photos(4)

Key Documents

SAB2108746

Sigma-Aldrich

Anti-MLKL

affinity isolated antibody

Synonym(s):

Anti-FLJ34389

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

54 kDa

species reactivity

human

concentration

0.5-1 mg/mL

technique(s)

immunoblotting: suitable

accession no.

NM_152649

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MLKL(197259)

General description

Mixed lineage kinase domain-like protein (MLKL) is encoded by the gene mapped to human chromosome 16q23. Activated MLKL is localized on the cell membrane.

Immunogen

Synthetic peptide directed towards the N terminal region of human MLKL

Biochem/physiol Actions

Mixed lineage kinase domain-like protein (MLKL) primarily causes receptor-interacting protein (RIP) kinase–dependent necroptosis. However, during hepatitis, it results in programmed hepatocellular necrosis which is independent of RIPK3.s MLKL also participates in endosomal trafficking and in the formation of extracellular vesicles.

Sequence

Synthetic peptide located within the following region: DVWKELSLLLQVEQRMPVSPISQGASWAQEDQQDADEDRRAFQMLRRDNE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

MLKL Activation Triggers NLRP3-Mediated Processing and Release of IL-1? Independently of Gasdermin-D.
Gutierrez KD
Journal of Immunology, 198, 2156-2156 (2017)
The pseudokinase MLKL mediates programmed hepatocellular necrosis independently of RIPK3 during hepatitis.
Gunther C
The Journal of Clinical Investigation, 126, 4346-4346 (2016)
MLKL, the Protein that Mediates Necroptosis, Also Regulates Endosomal Trafficking and Extracellular Vesicle Generation.
Yoon S
Immunity, 47, 51-51 (2017)
Genetic changes associated with testicular cancer susceptibility.
Pyle LC and Nathanson KL
Seminars in Oncology, 43, 575-575 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service