Skip to Content
Merck
All Photos(4)

Key Documents

SAB2108448

Sigma-Aldrich

Anti-PCNA

IgG fraction of antiserum

Synonym(s):

Anti-AIT

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

29 kDa

species reactivity

bovine, rat, dog, mouse, human

concentration

0.5-1 mg/mL

technique(s)

immunoblotting: suitable
immunohistochemistry: suitable

accession no.

NM_002592

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCNA(5111)

General description

Proliferating cell nuclear antigen (PCNA), a nuclear protein, is expressed ubiquitously in mammals. This homotrimer protein is a doughnut-shaped molecule that is expressed at high levels in the thymus, bone marrow, fetal liver, and few cells of the small intestine and colon. PCNA is mainly located in the nucleus. The PCNA gene is a single-copy gene mapped to human chromosome 20p13.

Immunogen

Synthetic peptide directed towards the C terminal region of human PCNA

Application

Anti-PCNA has been used in:
  • immunoblot (1:500) (1:1,000) (1:750)
  • far-western analysis (1:1000)
  • immunohistochemistry (1?:?50) (1:1000)
  • proliferating cell nuclear antigen (PCNA) overlay assay (1:1,000)

Biochem/physiol Actions

Proliferating cell nuclear antigen (PCNA) participates in DNA replication and repair. It acts as an auxiliary factor of polymerase δ. It plays a key role in the maturation of Okazaki fragments. It can be used as a prognostic and diagnostic marker in chronic lymphoid leukemia (CLL). High expression of PCNA protein is seen in breast and duodenal cancers.

Sequence

Synthetic peptide located within the following region: LNFFTKATPLSSTVTLSMSADVPLVVEYKIADMGHLKYYLAPKIEDEEGS

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gang Li et al.
Journal of cellular physiology (2019-02-26)
Renal cell carcinoma (RCC) is a common urinary system cancer with high morbidity and mortality rate. Clear cell renal cell carcinoma (ccRCC) is a highly aggressive and common type of RCC. More and effective therapeutic targets are badly needed for
Amaia González-Magaña et al.
Biomolecules, 10(4) (2020-04-12)
: Proliferating cell nuclear antigen (PCNA) is an essential factor in DNA replication and repair. It forms a homotrimeric ring that embraces the DNA and slides along it, anchoring DNA polymerases and other DNA editing enzymes. It also interacts with
Elsayed I Salim et al.
Asian Pacific journal of cancer prevention : APJCP, 17(3), 1023-1035 (2016-04-05)
The purpose of this study was to investigate the role of colon cancer stem cells (CSCs) during chemicallyinduced rat multi-step colon carcinogenesis with or without the treatment with a specific cyclooxygenase-2 inhibitor drug (celecoxib). Two experiments were performed, the first
Xiaolan Ye et al.
Medicine, 99(16), e19755-e19755 (2020-04-22)
Although proliferating cell nuclear antigen (PCNA) plays an important role in tumor proliferation and its expression level is closely related to the biological activity of tumor cells, PCNA expression in non-small cell lung cancer (NSCLC) has been seldom reported. In
Weixin Hou et al.
Journal of inflammation research, 14, 7295-7313 (2022-01-08)
Acute-on-chronic liver failure (ACLF) is a critical disease with a high fatality rate. Immune dysfunction and inflammatory responses are key risk factors in ACLF. Pyroptosis is a form of programmed cell death characterized by the release of inflammatory cytokines, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service