Skip to Content
Merck

SAB1410179

Anti-TREX1 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

AGS1, AGS5, CRV, DKFZp434J0310, DRN3, HERNS

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 38.9 kDa

species reactivity

human, mouse

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TREX1(11277)

General description

This gene encodes the major 3′->5′ DNA exonuclease in human cells. The protein is a non-processive exonuclease that may serve a proofreading function for a human DNA polymerase. It is also a component of the SET complex, and acts to rapidly degrade 3′ ends of nicked DNA during granzyme A-mediated cell death. Mutations in this gene result in Aicardi-Goutieres syndrome, chilblain lupus, and Cree encephalitis. Multiple transcript variants encoding different isoforms have been found for this gene. (provided by RefSeq)

Immunogen

TREX1 (NP_057465.1, 1 a.a. ~ 369 a.a) full-length human protein.

Sequence
MGPGARRQGRIVQGRPEMCFCPPPTPLPPLRILTLGTHTPTPCSSPGSAAGTYPTMGSQALPPGPMQTLIFFDMEATGLPFSQPKVTELCLLAVHRCALESPPTSQGPPPTVPPPPRVVDKLSLCVAPGKACSPAASEITGLSTAVLAAHGRQCFDDNLANLLLAFLRRQPQPWCLVAHNGDRYDFPLLQAELAMLGLTSALDGAFCVDSITALKALERASSPSEHGPRKSYSLGSIYTRLYGQSPPDSHTAEGDVLALLSICQWRPQALLRWVDAHARPFGTIRPMYGVTASARTKPRPSAVTTTAHLATTRNTSPSLGESRGTKDLPPVKDPGALSREGLLAPLGLLAILTLAVATLYGLSLATPGE

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library