Skip to Content
Merck
All Photos(1)

Key Documents

SAB1404764

Sigma-Aldrich

Monoclonal Anti-CNAP1 antibody produced in mouse

clone 4C9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

CAP-D2, CNAP1, KIAA0159, hCAP-D2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4C9, monoclonal

form

buffered aqueous solution

mol wt

antigen ~37 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NCAPD2(9918)

Immunogen

CNAP1 (AAH28182, 1240 a.a. ~ 1339 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
LRKMLDNFDCFGDKLSDESIFSAFLSVVGKLRRGAKPEGKAIIDEFEQKLRACHTRGLDGIKELEIGQAGSQRAPSAKKPSSGSRYQPLASTASDNDFVT

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Philipp Haselmayer et al.
Immunology, 119(3), 404-411 (2006-10-28)
In microbial infections polymorphnuclear neutrophils (PMN) constitute a major part of the innate host defence, based upon their ability to rapidly accumulate in inflamed tissues and clear the site of infection from microbial pathogens by their potent effector mechanisms. The
Johan Busselez et al.
Scientific reports, 9(1), 7187-7187 (2019-05-12)
We have used cryo Electron Tomography, proteomics and immunolabeling to study centrosomes isolated from the young lamb thymus, an efficient source of quiescent differentiated cells. We compared the proteome of thymocyte centrosomes to data published for KE37 cells, focusing on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service