Skip to Content
Merck
All Photos(3)

Key Documents

SAB1401004

Sigma-Aldrich

Anti-ADAR antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

ADAR1, DRADA, DSH, DSRAD, G1P1, IFI-4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADAR(103)

General description

Adenosine deaminase acting on RNA (ADAR) protein is an RNA -specific C-6 adenosine deaminase. This protein is composed of one or two N-terminal Z-DNA binding domains, three centrally located double-stranded RNA (dsRNA) binding domains, and a C-terminal deaminase catalytic domain. This protein has two isoforms present in several human cell lines. The ADAR gene is mapped on the human chromosome at 1q21.3.
This gene encodes the enzyme responsible for RNA editing by site-specific deamination of adenosines. This enzyme destabilizes double stranded RNA through conversion of adenosine to inosine. Mutations in this gene have been associated with dyschromatosis symmetrica hereditaria. Alternate transcriptional splice variants, encoding different isoforms, have been characterized. (provided by RefSeq)

Immunogen

ADAR (P55265, 1 a.a. ~ 1226 a.a) full-length human protein.

Sequence
MNPRQGYSLSGYYTHPFQGYEHRQLRYQQPGPGSSPSSFLLKQIEFLKGQLPEAPVIGKQTPSLPPSLPGLRPRFPVLLASSTRGRQVDIRGVPRGVHLGSQGLQRGFQHPSPRGRSLPQRGVDCLSSHFQELSIYQDQEQRILKFLEELGEGKATTAHDLSGKLGTPKKEINRVLYSLAKKGKLQKEAGTPPLWKIAVSTQAWNQHSGVVRPDGHSQGAPNSDPSLEPEDRNSTSVSEDLLEPFIAVSAQAWNQHSGVVRPDSHSQGSPNSDPGLEPEDSNSTSALEDPLEFLDMAEIKEKICDYLFNVSDSSALNLAKNIGLTKARDINAVLIDMERQGDVYRQGTTPPIWHLTDKKRERMQIKRNTNSVPETAPAAIPETRRNAEFLTCNIPTSNASNNMVTTEKVENGQEPVIKLENRQEARPEPARLKPPVHYNGPSKAGYVDFENGQWATDDIPDDLNSIRAAPGEFRAIMEMPSFYSHGLPRCSPYKKLTECQLKNPISGLLEYAQFASQTCEFNMIEQSGPPHEPRFKFQVVINGREFPPAEAGSKKVAKQDAAMKAMTILLEEAKAKDSGKSEESSHYSTEKESEKTAESQTPTPSATSFFSGKSPVTTLLECMHKLGNSCEFRLLSKEGPAHEPKFQYCVAVGAQTFPSVSAPSKKVAKQMAAEEAMKALHGEATNSMASDNQPEGMISESLDNLESMMPNKVRKIGELVRYLNTNPVGGLLEYARSHGFAAEFKLVDQSGPPHEPKFVYQAKVGGRWFPAVCAHSKKQGKQEAADAALRVLIGENEKAERMGFTEVTPVTGASLRRTMLLLSRSPEAQPKTLPLTGSTFHDQIAMLSHRCFNTLTNSFQPSLLGRKILAAIIMKKDSEDMGVVVSLGTGNRCVKGDSLSLKGETVNDCHAEIISRRGFIRFLYSELMKYNSQTAKDSIFEPAKGGEKLQIKKTVSFHLYISTAPCGDGALFDKSCSDRAMESTESRHYPVFENPKQGKLRTKVENGEGTIPVESSDIVPTWDGIRLGERLRTMSCSDKILRWNVLGLQGALLTHFLQPIYLKSVTLGYLFSQGHLTRAICCRVTRDGSAFEDGLRHPFIVNHPKVGRVSIYDSKRQSGKTKETSVNWCLADGYDLEILDGTRGTVDGPRNELSRVSKKNIFLLFKKLCSFRYRRDLLRLSYGEAKKAARDYETAKNYFKKGLKDMGYGNWISKPQEEKNFYLCPV

Application

Anti-ADAR antibody produced in rabbit has been used in immunoprecipitation.

Biochem/physiol Actions

Adenosine deaminase acting on RNA (ADAR) protein facilitates the conversion of adenosine to inosine in double-stranded RNAs (dsRNA). This protein is involved in the RNA interference pathway, to change the targeting and processing of microRNAs. Mutations in the ADAR gene lead to an auto-immune Aicardi-Goutières syndrome (AGS) and dyschromatosis symmetrica hereditarian (DSH).

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C X George et al.
Gene, 229(1-2), 203-213 (1999-03-30)
The double-stranded RNA-specific adenosine deaminase (ADAR1) is inducible by interferon (IFN) and is implicated in the editing of viral RNAs during lytic and persistent infection. We have now isolated and characterized human genomic clones that contain the promoter region required
Ann M Toth et al.
The Journal of biological chemistry, 284(43), 29350-29356 (2009-08-28)
ADAR1 (adenosine deaminase acting on RNA) catalyzes the conversion of adenosine to inosine, a process known as A-to-I editing. Extensive A-to-I editing has been described in viral RNAs isolated from the brains of patients persistently infected with measles virus, although
Cora Cravero et al.
Case reports in psychiatry, 2017, 7582780-7582780 (2017-06-20)
We report the case of a young boy with nonverbal autism and intellectual disability, with a rare de novo 1q21.3 microdeletion. The patient had early and extreme self-injurious behaviours that led to blindness, complicated by severe developmental regression. A significant
Noriyuki Suzuki et al.
The Journal of investigative dermatology, 124(6), 1186-1192 (2005-06-16)
Dyschromatosis symmetrica hereditaria (DSH) (also called "reticulate acropigmentation of Dohi") is a pigmentary genodermatosis of autosomal dominant inheritance. We have clarified for the first time four pathological mutations of the double-stranded RNA-specific adenosine deaminase gene (ADAR1 or DSRAD) in four
Gillian I Rice et al.
Nature genetics, 44(11), 1243-1248 (2012-09-25)
Adenosine deaminases acting on RNA (ADARs) catalyze the hydrolytic deamination of adenosine to inosine in double-stranded RNA (dsRNA) and thereby potentially alter the information content and structure of cellular RNAs. Notably, although the overwhelming majority of such editing events occur

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service