Skip to Content
Merck
All Photos(10)

Key Documents

SAB1400160

Sigma-Aldrich

Monoclonal Anti-SMAD3 antibody produced in mouse

clone 7F3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DKFZP586N0721, Anti-DKFZp686J10186, Anti-HSPC193, Anti-HsT17436, Anti-JV152, Anti-MADH3, Anti-MGC60396, Anti-Smad 3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7F3, monoclonal

form

buffered aqueous solution

species reactivity

human, rat

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
proximity ligation assay: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

UniProt accession no.

application(s)

research pathology

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMAD3(4088)

General description

Mothers against decapentaplegic homolog 3 (SMAD3) protein is an important constituent of the transforming growth factor-beta (TGF-β) signaling pathway. Mad homology 1 (MH1) and Mad homology 2 (MH2) are the two functional domains of the SMAD3. The SMAD3 gene is located on human chromosome 15q22.33.

Immunogen

SMAD3 (NP_005893, 120 a.a. ~ 221 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
VCVNPYHYQRVETPVLPPVLVPRHTEIPAEFPPLDDYSHSIPENTNFPAGIEPQSNIPETPPPGYLSEDGETSDHQMNHSMDAGSPNLSPNPMSPAHNNLDL

Application

Monoclonal Anti-SMAD3 antibody produced in mouse has been used in immunohistochemistry (1:200).

Biochem/physiol Actions

Mothers against decapentaplegic homolog 3 (SMAD3) protein possesses tumor-suppressive actions. It is involved in epithelial to mesenchymal transition (EMT). SMAD3 also possesses tumor-promoting actions of transforming growth factor-beta (TGF-β). The N-terminal domain Mad homology 1 (MH1) of SMAD3 plays a role in DNA binding. The SMAD3 gene mutation is linked to an increased incidence of knee osteoarthritis. DNA methylation change in the SMAD3 gene promoter is associated with atopic asthma. SMAD3 protein plays a regulatory role in immune responses. It also modulates fibrosis in the airways.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda C Daly et al.
The Journal of biological chemistry, 285(9), 6489-6497 (2009-12-29)
Transforming growth factor beta (TGF-beta) regulates many biological processes, and aberrant TGF-beta signaling is implicated in tumor development. Smad3 is a central component of the TGF-beta signaling pathway, and once activated, Smad3 forms complexes with Smad4 or other receptor-regulated Smads
Astrid Jeibmann et al.
Journal of neuro-oncology, 131(3), 477-484 (2017-01-22)
Atypical teratoid/rhabdoid tumors (ATRT) are highly malignant brain tumors arising in young children. The majority of ATRT is characterized by inactivation of the chromatin remodeling complex member SMARCB1 (INI1/hSNF5). Little is known, however, on downstream pathways involved in the detrimental
Bertrand Chesneau et al.
Molecular genetics & genomic medicine, 8(5), e1132-e1132 (2020-03-11)
Pathogenic SMAD3 variants are responsible for a cardiovascular phenotype, mainly thoracic aortic aneurysms and dissections. Precocious identification of the vascular risk such as aortic dilatation in mutated patients has a major impact in terms of management, particularly to avoid dissection
R J Lund et al.
Allergy, 73(8), 1735-1740 (2018-05-08)
Children with rhinovirus-induced severe early wheezing have an increased risk of developing asthma later in life. The exact molecular mechanisms for this association are still mostly unknown. To identify potential changes in the transcriptional and epigenetic regulation in rhinovirus-associated atopic
Chao Lu et al.
Molecular biology reports, 46(4), 4501-4505 (2019-06-12)
The mechanism of knee osteoarthritis (OA) is still not clearly elucidated. SMAD3 gene polymorphisms are considered to play a vital role in OA pathogenesis. We thus investigated the relationship of SMAD3 rs1065080 gene polymorphism and susceptibility to knee osteoarthritis in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service