Skip to Content
Merck
All Photos(4)

Documents

SAB1400151

Sigma-Aldrich

Anti-LBR antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-DHCR14B, Anti-LMN2R, Anti-MGC9041, Anti-PHA, Anti-PRO0650

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect immunofluorescence: suitable
western blot: 1 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LBR(3930)

General description

Lamin B receptor (LBR) is a transmembrane protein which is expressed in the inner nuclear membrane. It possesses an amino-terminal domain, eight transmembrane domains and a carboxyl-terminal region. The gene encoding LBR is localized on human chromosome 1q42.

Immunogen

LBR (NP_002287.2, 1 a.a. ~ 615 a.a) full-length human protein.

Sequence
MPSRKFADGEVVRGRWPGSSLYYEVEILSHDSTSQLYTVKYKDGTELELKENDIKPLTSFRQRKGGSTSSSPSRRRGSRSRSRSRSPGRPPKSARRSASASHQADIKEARREVEVKLTPLILKPFGNSISRYNGEPEHIERNDAPHKNTQEKFSLSQESSYIATQYSLRPRREEVKLKEIDSKEEKYVAKELAVRTFEVTPIRAKDLEFGGVPGVFLIMFGLPVFLFLLLLMCKQKDPSLLNFPPPLPALYELWETRVFGVYLLWFLIQVLFYLLPIGKVVEGTPLIDGRRLKYRLNGFYAFILTSAVIGTSLFQGVEFHYVYSHFLQFALAATVFCVVLSVYLYMRSLKAPRNDLSPASSGNAVYDFFIGRELNPRIGTFDLKYFCELRPGLIGWVVINLVMLLAEMKIQDRAVPSLAMILVNSFQLLYVVDALWNEEALLTTMDIIHDGFGFMLAFGDLVWVPFIYSFQAFYLVSHPNEVSWPMASLIIVLKLCGYVIFRGANSQKNAFRKNPSDPKLAHLKTIHTSTGKNLLVSGWWGFVRHPNYLGDLIMALAWSLPCGFNHILPYFYIIYFTMLLVHREARDEYHCKKKYGVAWEKYCQRVPYRIFPYIY

Biochem/physiol Actions

Lamin B receptor (LBR) inhibits immature chromatin binding of proteins. It has a role in the construction of the nuclear envelope in vitro and in vivo. Mutation in the gene encoding LBR has been linked to Pelger-Huët anomaly (PHA).

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Temporal control of nuclear envelope assembly by phosphorylation of lamin B receptor.
Tseng LC and Chen RH
Molecular Biology of the Cell (2011)
Dosage effect of zero to three functional LBRgenes
in vivo and in vitro
Sophia Gravemann
Nucleus (Austin, Tex.) (2010)
Shiwei Liu et al.
Nature, 561(7724), 551-555 (2018-09-21)
Defects in the architecture or integrity of the nuclear envelope are associated with a variety of human diseases1. Micronuclei, one common nuclear aberration, are an origin for chromothripsis2, a catastrophic mutational process that is commonly observed in cancer3-5. Chromothripsis occurs
An in vitro model for Pelger-Huet anomaly: stable knockdown of lamin B receptor in HL-60 cells.
Olins AL
Nucleus (Austin, Tex.) (2010)
Shangming Tang et al.
Nature, 606(7916), 930-936 (2022-04-28)
Chromothripsis is a catastrophic mutational process that promotes tumorigenesis and causes congenital disease1-4. Chromothripsis originates from aberrations of nuclei called micronuclei or chromosome bridges5-8. These structures are associated with fragile nuclear envelopes that spontaneously rupture9,10, leading to DNA damage when

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service