Skip to Content
Merck
All Photos(1)

Documents

S9555

Sigma-Aldrich

Anti-E-Selectin (CD62E) antibody, Mouse monoclonal

clone 1.2B6, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-CD62E, Anti-E-Selectin, Anti-ELAM-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1.2B6, monoclonal

form

buffered aqueous solution

mol wt

antigen 100-115 kDa

species reactivity

human, pig

concentration

~2 mg/mL

technique(s)

capture ELISA: suitable
flow cytometry: suitable
immunohistochemistry (frozen sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
indirect immunofluorescence: 1-2 μg/mL using using 293-T cells transfected with E-selectin fusion protein
western blot: suitable (non-reduced)

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SELE(6401)

Looking for similar products? Visit Product Comparison Guide

General description

Monoclonal anti-E-selectin (CD62E) (mouse IgG1 isotype) is derived from the 1.2B6 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with TNF-α activated human endothelial cells. This antibody reacts specifically with E-selectin (CD62E) expressed on the surface of activated endothelial cells. The antibody may weakly cross-react with recombinant human P-selectin and human platelet P-selectin. Cross-reactivity has been observed with porcine E-selectin.
It is a 140KDa protein mainly expressed in endothelial cells. E- selectin is characterized with an N-terminal domain, epidermal growth factor (EGF)-like domain, lectin (ligand-binding) domain, variable short consensus repeats and transmembrane domain.
Selectin E (SELE) is encoded by the gene mapped to human chromosome 1q24.2. The encoded protein is mainly expressed on luminal surfaces of inflamed vasculature.

Specificity

Reacts specifically with human CD62E (E-Selectin, ELAM-1) expressed on the surface of activated endothelial cells. The antibody may weakly cross-react with recombinant hman P-Selectin and human platelet P-Selectin. Cross-reactivity has been observed with porcine E-Selectin. Monoclonal Anti-human E-Selctin inhibits adhesion of neutrophils, eosinophils, and a skin-homing T-cell subpopulation to activated endothelial cells and CD62E-transfected cells. 5th Workshop: code no. E 040.

Immunogen

TNF-α activated human endothelial cells.

Application

Anti-E-Selectin (CD62E) antibody, Mouse monoclonal has been used in immunofluorescence.

Biochem/physiol Actions

E-selectin acts as a key inflammatory cytokine. E-selectin-dependent rolling helps in binding pancreatic cancer cell to hyaluronic acid. In addition, the encoded protein in association with P-selectins, plays a vital role in recruiting leukocytes to inflammatory sites. Mutation in the gene is associated with the development of various diseases such as atherosclerosis and stroke.
Selectins play a vital role in maintaining homoeostasis and cutaneous inflammation. SEL-E and SEL-P help in carrying T helper (Th1) cells into inflamed tissues. SEL-E acts as a receptor for cluster of differentiation 44 (CD44 (HCELL)), Mac-2 binding protein, mucin 1 (Muc-1), death receptor-3 and E-selectin ligand-1 (ESL-1). Overexpression of the gene might affect adhesion and migration of the cells leading to the endometriosis disorder.

Target description

CD62E (E-Selectin) surface antigen is a glycoprotein and member of the selectin family of adhesion receptors. It is expressed by activated human endothelial cells and is detectable in the plasma.

Physical form

Solution, 0.2 μm filtered, in 0.01 M phosphate buffered saline, pH 7.4.

Storage and Stability

For continuous use, store sterile at 2-8°C. For extended storage freeze in sterile working aliquots. Repeated freezing and thawing, or storage in "frostfree" freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Effect of C1-INH on ischemia/reperfusion injury in a porcine limb ex vivo perfusion model.
Abdelhafez MM, et al.
Molecular Immunology, 88, 116-124 (2017)
No association between E- and L-selectin genes and SLE: soluble L-selectin levels do correlate with genotype and a subset in SLE
Russell AI, et al.
Genes and Immunity, 6, 422-429 (2005)
Association of E-selectin Gene Polymorphism (S128R) with Ischemic Stroke and Stroke Subtypes
Roy S, et al.
Inflammation, 37, 599-603 (2014)
Burcin Ekser et al.
Transplant international : official journal of the European Society for Organ Transplantation, 25(8), 882-896 (2012-05-31)
Upregulation of tissue factor (TF) expression on activated donor endothelial cells (ECs) triggered by the immune response (IR) has been considered the main initiator of consumptive coagulopathy (CC). In this study, we aimed to identify potential factors in the development
E-selectin-mediated rolling facilitates pancreatic cancer cell adhesion to hyaluronic acid.
Shea DJ, et al.
Faseb Journal, 31, 5078-5086 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service