Skip to Content
Merck
All Photos(1)

Key Documents

S8826

Sigma-Aldrich

Semax

≥98% (HPLC)

Synonym(s):

ACTH (4-7), Pro-Gly-Pro trifluoroacetate salt, L-Methionyl-L-a-glutamyl-L-histidyl-L-phenylalanyl-L-prolylglycyl-L-proline trifluoroacetate salt, Met-Glu-His-Phe-Pro-Gly-Pro

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H51N9O10S · xC2HF3O2
CAS Number:
Molecular Weight:
813.92 (free base basis)
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

H2O: >2 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC(=O)C(F)(F)F.CSCC[C@H](N)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](Cc1c[nH]cn1)C(=O)N[C@@H](Cc2ccccc2)C(=O)N3CCC[C@H]3C(=O)NCC(=O)N4CCC[C@H]4C(O)=O

InChI

1S/C37H51N9O10S.C2HF3O2/c1-57-16-13-24(38)32(50)42-25(11-12-31(48)49)33(51)43-26(18-23-19-39-21-41-23)34(52)44-27(17-22-7-3-2-4-8-22)36(54)46-15-5-9-28(46)35(53)40-20-30(47)45-14-6-10-29(45)37(55)56;3-2(4,5)1(6)7/h2-4,7-8,19,21,24-29H,5-6,9-18,20,38H2,1H3,(H,39,41)(H,40,53)(H,42,50)(H,43,51)(H,44,52)(H,48,49)(H,55,56);(H,6,7)/t24-,25-,26-,27-,28-,29-;/m0./s1

InChI key

UAXRUQRHZBYPHF-IGCYJKAMSA-N

General description

Semax regulates the expression and the activation of the hippocampal (brain-derived neurotrophic factor) BDNF/(tropomyosin receptor kinase B) trkB system. It enhances selective attention at the time of information reception, increases memory consolidation and stimulates learning abilities. Semax is an active element of SEMAX drugs, which is used for the treatment of brain hypoxia and ischemia. It stimulates the activity of choline acetyltransferase. Semax exhibits properties of the melanocortin (MC) receptor antagonist.

Biochem/physiol Actions

Semax (Met-Glu-His-Phe-Pro-Gly-Pro) is an analog of the adrenocorticotropin fragment (4-10). It activates the transcription of neurotrophins and has neuroprotective and nootropic activity.
Semax is a neuroprotective ACTH (4-10) analog.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D M Manchenko et al.
Rossiiskii fiziologicheskii zhurnal imeni I.M. Sechenova, 96(10), 1014-1023 (2011-01-29)
Heptapeptide Semax (MEHFPGP) is the fragment of ACTH(4-10) analogue with prolonged neurotropic activity. The aim of the present work was to study the Semax effects on learning capability and pain sensitivity in white rats following intraperitoneal and intranasal administration in
Vasily V Stavchansky et al.
Journal of molecular neuroscience : MN, 45(2), 177-185 (2010-07-10)
The neuropeptide preparation Semax (Met-Glu-His-Phe-Pro-Gly-Pro) has been employed successfully in clinical practice for treating patients with severe brain blood circulation disorders. In spite of numerous studies, many aspects of the therapeutic effects of this preparation remain unknown. In this context
G A Romanova et al.
Patologicheskaia fiziologiia i eksperimental'naia terapiia, (3)(3), 62-67 (2012-10-18)
It was stated, that with bilateral photochemically induced thrombosis of the prefrontal cortex peptide semax and the AB-Glu by intranasal injection provoke pronounced neuroprotective and antiamnestic action. Intranasal injection semax (250 mkg/kg/daily during six postoperative days) and AB-Glu (250 mkg/kg
Ex vivo screening of prospective peptide drugs: new approaches.
L A Andreeva et al.
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections, 434, 300-303 (2010-10-22)
V V Stavchanskiĭ et al.
Molekuliarnaia biologiia, 45(6), 1026-1035 (2012-02-03)
Neurotrophins regulate key function of nervous tissue cells. Analysis of neurotrophins mRNA expression is an appropriate tool to assess therapeutic efficiency of the anti-stroke drugs. We have analyzed the effect of synthetic peptide semax and its C-terminal Pro-Gly-Pro tripeptide upon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service