Skip to Content
Merck

S8295

Sodium selenate

BioXtra

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
Na2SeO4
CAS Number:
Molecular Weight:
188.94
NACRES:
NA.21
PubChem Substance ID:
UNSPSC Code:
12352300
EC Number:
236-501-8
MDL number:
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioXtra

Quality Segment

form

powder

impurities

≤0.01% Phosphorus (P), ≤0.1% Insoluble matter

pH

5.5-7.5 (25 °C, 18.9 g/L)

anion traces

chloride (Cl-): ≤0.05%, sulfate (SO42-): ≤0.1%

cation traces

Al: ≤0.0005%, Ca: ≤0.005%, Cu: ≤0.0005%, Fe: ≤0.0005%, K: ≤0.02%, Mg: ≤0.0005%, NH4+: ≤0.05%, Pb: ≤0.001%, Zn: ≤0.0005%

SMILES string

[Na+].[Na+].[O-][Se]([O-])(=O)=O

InChI

1S/2Na.H2O4Se/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

MHQOTKLEMKRJIR-UHFFFAOYSA-L

Application

Sodium selenate has been used as an inorganic source of selenium:
  • to study its effect on the immune response and disease resistance of prawn
  • to study its antioxidant defense and in glucose homeostasis in streptozotocin-induced diabetes
  • as standard in enzymatic hydrolysis



Still not finding the right product?

Explore all of our products under Sodium selenate


signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - STOT RE 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library