Skip to Content
Merck

S8064

Solvent Safe pipette tips

volume range 100-1000 μL, sterile

Synonym(s):

carbon fiber pipette tips, solvent safe pipette tips, MBP 5479E, dispensing tip, liquid transfer tip

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.01
UNSPSC Code:
41121601
Volume:
100—1000 μL
Sterility:
sterile
Feature:
barrier
Packaging:
pack of 400 ea (4 trays of 100 ea)
Manufacturer/tradename:
Molecular BioProducts 5479E
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

carbon fiber , colorless

sterility

sterile

feature

barrier

packaging

pack of 400 ea (4 trays of 100 ea)

manufacturer/tradename

Molecular BioProducts 5479E

L

3.5 in. (88.3 mm)

maximum volume

1 mL

volume range

100-1000 μL

volume

100—1000 μL

compatibility

for use with

  • Biohit Variable 200-1000 μl
  • Eppendorf Research Pro 50-1000 μl
  • Eppendorf Response 4850 50-1000 μl
  • Finnpipette 4027/4500 Digital 200-1000 μl
  • Nichiryo BenchMate 100-1000 μl
  • Pipetman P-1000 100-1000 μl
  • Rainin EDP-Series 1000 100-1000 μl
  • Socorex/Wheaton 822 Series 100-1000 μl
  • Transferpette 100-1000 μl

General description

Solvent Safepipette rips are suitable for use with acids, bases and aggressive organic solvents Revolutionary filter material handles the pipetting challenges of combinatorial chemistry. The unique use of the ART™ filter with pleated activated carbon provides protection for your samples and pipettors against destructive carryover aerosols and vapors from volatile organic solvents without compromising pipetting accuracy and precision.

Application

  • Peptide synthesis
  • HPLC analysis

Features and Benefits

  • Solvent Safe carbon filter pipette tips
  • Uses the new ART filter tip formula (aerosol-resistant tip)
  • Prevents pipetting errors and critical inaccuracies caused by strong acids, bases and aggressive organic solvents
  • A proprietary blend of pleated activated carbon with a 40-micron polyethylene filter provides effective solvent protection
  • Filters block corrosive vapors created when pipetting volatile organic solvents such as TFA
  • Safely and accurately pipette phenol/chloroform DNA extractions
  • Perform peptide synthesis and HPLC analysis
  • Excellent alternative to glass pipettes

Other Notes

Browse all pipette tips

Legal Information

Solvent Safe is a trademark of Molecular BioProducts, Inc.


Still not finding the right product?

Explore all of our products under Solvent Safe pipette tips




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library