Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C8H14N2O5
CAS Number:
Molecular Weight:
218.21
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Product Name
Siastatin B, lyophilized powder, from microbial
biological source
microbial
Quality Segment
form
lyophilized powder
packaging
vial of 23.0 μmol
solubility
H2O: 2.3 mL/vial (for a 10 mM solution)
storage temp.
−20°C
SMILES string
CC(=O)N[C@H]1NC[C@@H]([C@H](O)[C@@H]1O)C(O)=O
InChI
1S/C8H14N2O5/c1-3(11)10-7-6(13)5(12)4(2-9-7)8(14)15/h4-7,9,12-13H,2H2,1H3,(H,10,11)(H,14,15)/t4-,5-,6-,7+/m0/s1
InChI key
DQTKLICLJUKNCG-ZTYPAOSTSA-N
Application
Broad spectrum inhibitor of sialidase.
Biochem/physiol Actions
Streptomyces metabolite, a broad-spectrum inhibitor of neuraminidase (sialidase).
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
