Skip to Content
Merck
All Photos(1)

Documents

S6881

Sigma-Aldrich

Syringic acid

≥95% (HPLC)

Synonym(s):

3,5-Dimethoxy-4-hydroxybenzoic acid, 4-Hydroxy-3,5-dimethoxy-benzoic acid, Gallic acid 3,5-dimethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H2(OCH3)2CO2H
CAS Number:
Molecular Weight:
198.17
Beilstein:
2115262
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (HPLC)

form

powder

mp

205-209 °C (lit.)

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

COc1cc(cc(OC)c1O)C(O)=O

InChI

1S/C9H10O5/c1-13-6-3-5(9(11)12)4-7(14-2)8(6)10/h3-4,10H,1-2H3,(H,11,12)

InChI key

JMSVCTWVEWCHDZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Effects of preheat treatment and syringic acid modification on the structure, functional properties, and stability of black soybean protein isolate.: This paper discusses how syringic acid modifications can enhance the functional properties and stability of black soybean protein, which is crucial for food science applications (Huang J et al., 2024).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah C Forester et al.
Journal of agricultural and food chemistry, 56(19), 9299-9304 (2008-09-05)
Anthocyanins are polyphenol antioxidants that have been shown to prevent many chronic diseases, including colon cancer. The compounds are largely metabolized by various enzymes and bacteria in the large intestine, and the health benefits of consuming foods rich in anthocyanins
Ayano Itoh et al.
Biological & pharmaceutical bulletin, 33(6), 983-987 (2010-06-05)
The mycelia of the edible mushroom Lentinula edodes can be cultured in solid medium containing lignin, and the hot-water extracts (L.E.M.) is commercially available as a nutritional supplement. During the cultivation, phenolic compounds, such as syringic acid and vanillic acid
Joanna Klepacka et al.
Plant foods for human nutrition (Dordrecht, Netherlands), 66(1), 64-69 (2011-01-19)
The aim of the study was to determine whether phenolic compounds in some varieties of buckwheat, winter and spring barley and peas can be used as factors which distinguish selected cultivars and varieties of plant material. It was observed that
Ilhami Gülçin et al.
Journal of food science, 76(4), C585-C593 (2012-03-16)
The raspberry (Rubus idaeus L.) is an economically important berry crop that contains many phenolic compounds with potential health benefits. In this study, important pomological features, including nutrient content and antioxidant properties, of a domesticated and 3 wild (Yayla, Yavuzlar
Doris M Jacobs et al.
Journal of agricultural and food chemistry, 60(12), 3078-3085 (2012-03-01)
Red wine and grape polyphenols are considered to promote cardiovascular health and are involved in multiple biological functions. Their overall impact on the human metabolome is not known. Therefore, exogenous and endogenous metabolic effects were determined in fasting plasma and

Protocols

Protocol for HPLC Analysis of Flavonoids on Ascentis® RP-Amide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service