Skip to Content
Merck
All Photos(1)

Documents

S6657

Sigma-Aldrich

Superdex®

Superdex®75 Prep Grade

Synonym(s):

Superdex Resin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
NACRES:
NA.56

form

suspension (20% aqueous ethanol)

technique(s)

protein array: suitable

matrix

cross-linked agarose

particle size

24-44 μm

pore size

3-70 kDa MW range (proteins)
500-30,000 Da MW range (dextrans)

operating pH range

1-14(short term)
3-12(long term)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Combines the high selectivity of a Sephadex matrix with the chemical and physical stability of crosslinked agarose.

Application

Superdex® is used for protein chromatography, gel filtration chromatography, gel filtration media, resins and separation media. Superdex® has been used for the purification and characterization of a novel laccase from the edible mushroom Hericium coralloides. Superdex® has also been used for the purification and mechanism exploration of a potential human hepatocellular carcinoma inhibitor from Bauhinia purpurea L. seeds.
Optimized for high resolution preparative separations by size exclusion chromatography. Recommended fractionation ranges are listed for globular proteins and dextrans.

Legal Information

Superdex is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F

Flash Point(C)

40 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cornelia Steinhauer et al.
Proteomics, 6(15), 4227-4234 (2006-07-11)
Antibody-based microarray is a novel technology with great promise in biomedicine that will provide unique means to perform global proteome analysis. In the process of designing the high-density antibody microarrays required, several critical key issues have been identified that remain
M Belew et al.
Journal of chromatography. A, 679(1), 67-83 (1994-09-09)
Recombinant human granulocyte-macrophage colony-stimulating factor (rhGM-CSF), produced as inclusion bodies in genetically transformed Escherichia coli cells was purified to homogeneity by a three-step chromatographic procedure involving hydrophobic interaction, ion exchange and gel filtration. Each purification step is reproducible and well
Toshiro Matsunaga et al.
Plant physiology, 134(1), 339-351 (2003-12-13)
Borate ester cross-linking of the cell wall pectic polysaccharide rhamnogalacturonan II (RG-II) is required for the growth and development of angiosperms and gymnosperms. Here, we report that the amounts of borate cross-linked RG-II present in the sporophyte primary walls of
Xindu Geng et al.
Journal of biotechnology, 113(1-3), 137-149 (2004-09-24)
A new technology for renaturation with simultaneous purification of the recombinant human interferon-gamma (rhIFN-gamma) in downstream of biotechnology is presented. The strategies to develop the new technology in industry scale were suggested. Based on chemical equilibrium and molecular interactions, the
S Gibert et al.
Journal of chromatography. B, Biomedical sciences and applications, 737(1-2), 143-150 (2000-02-19)
A kinesin gene has been cloned by RT-PCR (reverse transcription polymerase chain reaction) from Trypanosoma brucei and the corresponding protein overexpressed as a recombinant His-tag (histidine-tag) kinesin in E. coli in order to study its biochemical properties and to determine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service