Skip to Content
Merck

S5799

Suberic acid bis(3-sulfo-N-hydroxysuccinimide ester) sodium salt

≥95% (H-NMR), powder

Synonym(s):

BS3, Bis(sulfosuccinimidyl) suberate sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H18N2Na2O14S2
CAS Number:
Molecular Weight:
572.43
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352106
MDL number:
Assay:
≥95% (H-NMR)
Form:
powder
Solubility:
H2O: 10 mg/mL, soluble
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (H-NMR)

form

powder

reaction suitability

reagent type: cross-linking reagent

solubility

H2O: 10 mg/mL, soluble

storage temp.

−20°C

SMILES string

[Na+].[Na+].[O-]S(=O)(=O)C1CC(=O)N(OC(=O)CCCCCCC(=O)ON2C(=O)CC(C2=O)S([O-])(=O)=O)C1=O

InChI

1S/C16H20N2O14S2.2Na/c19-11-7-9(33(25,26)27)15(23)17(11)31-13(21)5-3-1-2-4-6-14(22)32-18-12(20)8-10(16(18)24)34(28,29)30;;/h9-10H,1-8H2,(H,25,26,27)(H,28,29,30);;/q;2*+1/p-2

InChI key

MGJYOHMBGJPESL-UHFFFAOYSA-L

General description

Suberic acid bis(3-sulfo-N-hydroxysuccinimide ester) sodium salt (BS3) is a water soluble, homobifunctional cross-linking reagent with amine reactivity. It is typically coupled to molecules containing primary amines by amide bonds buffered at pH 7.5 (6.5-8.5). It incorporates 8-atom linker.

Application

Suberic acid bis(3-sulfo-N-hydroxysuccinimide ester) sodium salt (BS3) is useful for the preparation of protein-protein, receptor-ligand and hapten carrier molecule conjugates.


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library