Skip to Content
Merck
All Photos(4)

Key Documents

S4197

Sigma-Aldrich

Anti-Sirt6 (C-terminal) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-NAD-dependent deacetylase sirtuin 6, Anti-SIR2-like protein 6, Anti-SIR2L6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~40 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

indirect immunofluorescence: suitable
western blot: 0.5-1 μg/mL using whole extracts of human U87 cells
western blot: 2.5-5 μg/mL using human HeLa cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SIRT6(51548)

General description

Sirtuin 6 (Sirt6) is a nuclear, chromatin-associated protein. Sirt6 is a member of Sir2 gene family. It is broadly expressed with the highest levels in muscle, brain and heart.

Immunogen

synthetic peptide corresponding to amino acids 330-348 of human Sirt6.

Application

Anti-Sirt6 (C-terminal) antibody has been used in
  • immunoblotting
  • immunofluorescence
  • western blotting
  • immunohistochemistry

Biochem/physiol Actions

Sirtuin 6 (Sirt6) plays a key role in DNA repair and maintenance of genomic stability in cells. Sirt6 is enzymatically inactive as a NAD+ -dependent protein deacetylase but displays auto-ADP-ribosyltransferase activity. Loss of Sirt6 leads to the development of an acute degenerative aging-like phenotype.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sirtuin 6 (SIRT6) rescues the decline of homologous recombination repair during replicative senescence
Mao Z, et al.
Proceedings of the National Academy of Sciences of the USA, 109(29), 11800-11805 (2012)
Sirtuin 6 expression and inflammatory activity in diabetic atherosclerotic plaques: effects of incretin treatment
Balestrieri M L, et al.
Diabetes, 64(4), 1395-1406 (2015)
The NAD+-dependent histone deacetylase SIRT6 promotes cytokine production and migration in pancreatic cancer cells by regulating Ca2+ responses
Bauer I, et al.
The Journal of Biological Chemistry, 287(49), 40924-40937 (2012)
Anti-inflammatory properties of sirtuin 6 in human umbilical vein endothelial cells
Lappas, Martha
Mediators of Inflammation, 2012 (2012)
Juan Ge et al.
Aging, 11(7), 1965-1976 (2019-03-31)
It has been well recognized that oocyte quality declines in aging animals. However, to date, the underlying mechanism remains to be explored. In the present study, we report that oocytes and embryos from aged mice (42-45 weeks old) display the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service