Skip to Content
Merck

S3772

Sheep serum

Synonym(s):

Serum, ovine serum

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.46
UNSPSC Code:
12352203
Biological source:
sheep
Sterility:
non-sterile
Shipped in:
dry ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

sheep

Quality Segment

conjugate

unconjugated

sterility

non-sterile

contains

preservative

shipped in

dry ice

storage temp.

−20°C

General description

Sheep Serum is a pooled preparation obtained from a normal donor herd.

Application

Sheep serum can be used as a blocking agent and control in immunoassays. It has also been used to block non-specific reactivity in immunohistochemistry of frozen rat levator labii superioris muscle tissue sections.
Sheep serum has been used as a blocking agent and control in immunoassays.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Explore all of our products under Sheep serum


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library