Skip to Content
Merck
All Photos(1)

Key Documents

S2890

Sigma-Aldrich

Streptavidin−Alkaline Phosphatase from Streptomyces avidinii

lyophilized powder, ≥700 DEA units/mg protein

Synonym(s):

Streptavidin-AP, Streptavidin−AP from Streptomyces avidinii

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

conjugate

alkaline phosphatase conjugate

Quality Level

form

lyophilized powder

specific activity

≥700 DEA units/mg protein

composition

Protein, ≥40% biuret

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Streptavidin, a 58 kDa protein, is a homotetramer purified from Streptomyces avidinii. It has a high affinity for biotin. Streptavidin has been conjugated with either peroxidase or alkaline phosphatase (ALP). These enzymes are used as protein conjugates because of their comparative stability and a wide range of suitable substrates for different applications. ALP conjugates to streptavidin in immunoassays.

Application

Streptavidin-alkaline phosphatase from Streptomyces avidinii has been used in enzyme-linked immune absorbent spot (ELISpot) to detect interferon (IFN-γ) producing cells.
The optimal working dilution should be determined empirically using a range of dilutions from a 1 mg/ml stock in buffer.

Unit Definition

One alkaline phosphatase unit will hydrolyze 1.0 μmole of p-nitrophenyl phosphate per min at pH 9.8 at 37 °C.

Physical form

Lyophilized powder containing trehalose with EPPS, phosphate and traces of magnesium chloride and zinc chloride.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of bovine Herpesvirus Type 5 on development of in vitro-produced bovine embryos.
Silva-Frade C
Theriogenology, 73(3), 324-331 (2010)
An allosteric-probe for detection of alkaline phosphatase activity and its application in immunoassay
Guo J, et al.
Frontiers in Chemistry, 6, 618-618 (2018)
Ola Blixt et al.
The Journal of biological chemistry, 278(33), 31007-31019 (2003-05-30)
Ten of the 11 known human siglecs or their murine orthologs have been evaluated for their specificity for over 25 synthetic sialosides representing most of the major sequences terminating carbohydrate groups of glycoproteins and glycolipids. Analysis has been performed using
Bystander activation of irrelevant CD4+ T cells following antigen-specific vaccination occurs in the presence and absence of adjuvant
van Aalst S, et al.
PLoS ONE, 12(5), e0177365-e0177365 (2017)
Microbial Resource Conservation: Conventional to Modern Approaches (2018)

Protocols

Enzymatic Assay of Alkaline Phosphatase, Diethanolamine Assay (EC 3. 1. 3. 1)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service