Skip to Content
Merck
All Photos(1)

Key Documents

S1639

Sigma-Aldrich

Complement sera from guinea pig

lyophilized powder

Synonym(s):

Complement serum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61
biological source:
guinea pig
form:
lyophilized powder
shipped in:
dry ice
Pricing and availability is not currently available.

biological source

guinea pig

Quality Level

form

lyophilized powder

technique(s)

cell culture | embryo: suitable

shipped in

dry ice

storage temp.

−20°C

Application

Complement sera from guinea pig has been used to incubate embryos for differential staining[1] and to stain DNA trophectoderm in embryo.[2]

Physical form

Lyophilized powder from indicated amount of serum

Analysis Note

Hemolytic titer (CH50 units per ml) is determined by method of Kabat and Mayer. Minimum 80 CH50 units per mL. Actual titer given on label.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Derivation of human embryonic stem cells in standard and chemically defined conditions.
Eric Chiao et al.
Methods in cell biology, 86, 1-14 (2008-04-30)
Yue Chen et al.
PLoS pathogens, 17(4), e1009535-e1009535 (2021-04-22)
The Peptidoglycan (PG) cell wall of the Lyme disease (LD) spirochete, Borrelia burgdorferi (Bb), contributes to structural and morphological integrity of Bb; is a persistent antigen in LD patients; and has a unique pentapeptide with L-Ornithine as the third amino
Involvement of histone H2B monoubiquitination in the regulation of mouse preimplantation development
Ooga M, et al.
Journal of Reproduction and Development, 2014-2137 (2015)
Guenhaël Sanz et al.
Biochimica et biophysica acta. General subjects, 1863(10), 1608-1618 (2019-07-07)
The placenta controls exchanges between the mother and the fetus and therefore fetal development and growth. The maternal environment can lead to disturbance of placental functions, with consequences on the health of the offspring. Since the rabbit placenta is very
Kerry J Laing et al.
Nature communications, 13(1), 6957-6957 (2022-11-15)
Herpes zoster is a localized skin infection caused by reactivation of latent varicella-zoster virus. Tissue-resident T cells likely control skin infections. Zoster provides a unique opportunity to determine if focal reinfection of human skin boosts local or disseminated antigen-specific tissue-resident

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service