Skip to Content
Merck
All Photos(1)

Key Documents

S1515

Sigma-Aldrich

Anti-Spectrin antibody produced in rabbit

whole antiserum

Synonym(s):

Anti-EL2, Anti-HPP, Anti-HS3, Anti-SPH3, Anti-SPTA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

human

technique(s)

western blot: 1:400

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Spectrin is a multifunctional protein that has lipid-binding sites within CH (calponin-homology) tandem domains, a PH (pleckstrin homology) domain and triple helical segments. It is located on chromosome 1q23.1.
Spectrin is the major cytoskeletal component in erythrocytes. It is composed of two types of polypeptides known as α and β with molecular weights of 240 and 220 kDa, respectively. The α- and β-chains form heterotetramers which are linked end-to-end via oligomeric actin and band 4.1 protein.

Specificity

The antibody reacts specifically with the α and β chains of human erythrocyte spectrin.

Immunogen

spectrin from freshly prepared human erythrocyte ghosts.

Application

Anti-Spectrin antibody has been used in western blotting immunofluorescence and immunostaining.

Biochem/physiol Actions

Spectrins modulates the cell morphology and mechanical properties. Spectrins also participates in the cell cycle by controlling the upregulation of membrane receptors. α- and β spectrins are essential for the development of nervous system.
The spectrin-based membrane skeleton is essential to provide mechanical stability and membrane integrity. It is also responsible for protein organization, trafficking and resilience of erythrocytes.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Choudhury et al.
Parasitology research, 83(8), 746-754 (1997-01-01)
Identification of neo-antigenic determinant(s) on parasite infected cell surface is important to control intracellular infections. Such determinant(s) on the surface of intact Plasmodium berghei infected erythrocytes have not been conclusively demonstrated. To generate polyclonal antiserum selectively recognizing the parasite infected
Chanjae Lee et al.
Development genes and evolution, 219(6), 319-330 (2009-06-26)
One contributing factor in the worldwide decline in amphibian populations is thought to be the exposure of eggs to UV light. Enrichment of pigment in the animal hemisphere of eggs laid in the sunlight defends against UV damage, but little
Natalie J Spillman et al.
Cellular microbiology, 19(6) (2017-01-10)
The malaria parasite exports numerous proteins into its host red blood cell (RBC). The trafficking of these exported effectors is complex. Proteins are first routed through the secretory system, into the parasitophorous vacuole (PV), a membranous compartment enclosing the parasite.
Physiologically important secondary modifications of red cell membrane in hereditary spherocytosis-evidence for in vivo oxidation and lipid rafts protein variations
Margetis P, et al.
Blood Cells, Molecules and Diseases, 38(3), 210-220 (2007)
F Galbiati et al.
Proceedings of the National Academy of Sciences of the United States of America, 97(17), 9689-9694 (2000-08-10)
It recently was reported that Duchenne muscular dystrophy (DMD) patients and mdx mice have elevated levels of caveolin-3 expression in their skeletal muscle. However, it remains unknown whether increased caveolin-3 levels in DMD patients contribute to the pathogenesis of DMD.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service