Skip to Content
Merck
All Photos(2)

Key Documents

S1006

Sigma-Aldrich

Sulfatides from bovine brain

≥97% (TLC), powder

Synonym(s):

Cerebroside sulfate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Sulfatides from bovine brain,

Quality Level

storage temp.

−20°C

General description

Sulfatideis a glycospingolipid and plays an essential role in host-pathogen recognition,β-cell function, infection, haemostasis/thrombosis, protein trafficking, cellgrowth, cell adhesion, neuronal plasticity, cell morphogenesis, and signaltransduction.[1][2] It is found on the surface of the blood cells such asplatelets, leukocytes, and erythrocytes. Sulfatides are also found in the serumas glycosphingolipids in lipoproteins.[3] It is made up of glycan part, a hydrophobic part, 3-O-sulfated galactose, and ceramide, which in turn is composed of a sphingosine base and a fatty acid.[1] It is an important component forthe maintenance of the myelin sheath andproper formation of the axo-glial junctions. Variations in the production ofsulfatides are associated with various human diseases like cancer andimmunological disorders. Alterations in the sulfatide metabolism lead to synapticdysfunction, neurological decline, inflammation, and myelin deterioration.[1]
Sulfatide is a 3-O-sulfogalactosylceramide consisting primarily of galacto cerebroside sulfate. It is localized in myelin sheath and comprises approximately 4% of total myelin lipid in brain.[4]

Application

Sulfatides has been used in:
  • enzyme linked immunosorbent assay (ELISA) in human plasma samples[5]
  • thin layer chromatography in myelin membranes[6]
  • in sulfatide binding assay of properdin[7]

Biochem/physiol Actions

Sulfatides play a key role in physiological processes of nervous system, immune system and in insulin secretion.[4] Anomalies in sulfatides catabolism in lysosome is implicated in late infantile metachromatic leukodystrophy.[8] High levels of sulfatides is also observed in ovarian cancer.[9] Sulfatides contribute to the pathogenesis of chronic inflammatory diseases of the central nervous system.[10]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalia A Crivello et al.
The Journal of nutritional biochemistry, 21(11), 1083-1088 (2010-01-23)
Dysregulation of myelin sulfatides is a risk factor for cognitive decline with age. Vitamin K is present in high concentrations in the brain and has been implicated in the regulation of sulfatide metabolism. Our objective was to investigate the age-related
Age-and brain region-specific effects of dietary vitamin K on myelin sulfatides
Crivello NA, et al.
The Journal of Nutritional Biochemistry, 21(11), 1083-1088 (2010)
Quantification of sulfatides and lysosulfatides in tissues and body fluids by liquid chromatography tandem mass spectrometry
Mirzaian M, et al.
Journal of Lipid Research, 56(8), 936-943 (2015)
Sulfatide-binding assay for von Willebrand factor: detection of von Willebrand's disease without discrimination of vWD subtypes
Favaloro EJ
Thrombosis Research, 98(2), 213-219 (2000)
Elevation of sulfatides in ovarian cancer: an integrated transcriptomic and lipidomic analysis including tissue-imaging mass spectrometry
Liu Y, et al.
Molecular Cancer, 9(1), 186-186 (2010)

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Questions

  1. Hi, Do you have any information about the sulfatide composition/profile of “Sulfatide from bovine brain (S1006)”.

    1 answer
    1. As reported in the 'DESCRIPTION' section of the product page, Sulfatide is a 3-O-sulfogalactosylceramide consisting primarily of galacto cerebroside sulfate. This product has not been further characterized.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service