Skip to Content
Merck

Skip To

S0692

Spectinomycin

Ready Made Solution, 100 mg/mL in DMSO/H2O, 1:1

Synonym(s):

Actinospectacin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
51281602
NACRES:
NA.85
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Streptomyces sp.

Quality Segment

form

ready-to-use solution

concentration

100 mg/mL in DMSO/H2O, 1:1

color

colorless

solubility

DMSO/H2O, 1:1: soluble 100 mg/mL

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria, mycobacteria

mode of action

protein synthesis | interferes

shipped in

wet ice

storage temp.

−20°C

General description

Chemical structure: aminoglycoside

Application

Spectinomycin is an aminocyclitol antibiotic derived from Streptomyces spectabilis. It has been shown to have a wide variety of uses, including treating acute gonorrheal urethritis and cervictis, to mark cell layers to monitor cell fate during leaf development, as a selection marker in plant related transformation systems for plant cells containing the marker gene Spcr, to study respiratory tract infections of cattle caused by Pasteurella multocida and Mannheimia haemolytica, and to generate plants deficient for the plastid-encoded RNA polymerase on MS-agar media. Spectinomycin also has use in amplification of low copy number plasmid carrying replicons. It is recommended for use in cell culture applications at 7.5-20 mg/L.

Biochem/physiol Actions

Mode of Action: Spectinomycin acts by inhibiting protein synthesis and elongation by binding to the bacterial 30S ribosomal subunit and interfering with peptidyl tRNA translocation.

Mode of Resistance: A mutation in rpsE, the gene for ribosomal protein S5, prevents the binding of spectinomycin. Resistance is also conferred by the aminoglycoside-3′-adenyltransferase gene (aadA).

Antimicrobial spectrum: Spectinomycin acts against gram-negative and gram-positive bacteria.

Preparation Note

Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.Store under inert gas.
Stock solutions should be prepared at 10 mg/mL in water and 0.2 μm filtered.

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store under inert gas

Disclaimer

Solutions can be stored at 2-8°C for several weeks or at -20°C for more extended periods.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S9007S2647S6501
description

Ready Made Solution, 100 mg/mL in DMSO/H2O, 1:1

description

potency: ≥603 μg per mg

description

powder, γ-irradiated

description

powder

mode of action

protein synthesis | interferes

mode of action

protein synthesis | interferes

mode of action

protein synthesis | interferes

mode of action

protein synthesis | interferes

antibiotic activity spectrum

mycobacteria, Gram-positive bacteria, Gram-negative bacteria

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

antibiotic activity spectrum

Gram-negative bacteria, Gram-positive bacteria

antibiotic activity spectrum

Gram-positive bacteria, Gram-negative bacteria, mycobacteria

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

ready-to-use solution

form

powder

form

powder

form

powder

solubility

DMSO/H2O, 1:1: soluble 100 mg/mL

solubility

H2O: soluble 50 mg/mL

solubility

H2O: soluble at 

solubility

-

concentration

100 mg/mL in DMSO/H2O, 1:1

concentration

-

concentration

-

concentration

-


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

201.0 °F - closed cup

flash_point_c

93.9 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–3 of 3 Questions  
  1. Can I use it for bacterial culture, since it has dmso?

    1 answer
    1. DMSO is typically safe to use in cell culture providing that the final concentration in medium does not exceed 0.5%. This product is a 100 mg/mL solution prepared in a 1:1 mixture of DMSO and water. The recommended concentration of this product for use in cell culture is 7.5 - 20 mg/L. At the maximum suggested concentration of 20 mg/L, the final concentration of DMSO is approximately 0.01%; well within tolerable limits for cells.

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  3. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters