Skip to Content
Merck
All Photos(2)

Key Documents

RDRT

Sigma-Aldrich

ReadyScript® cDNA Synthesis Mix

Complete reagent for first strand cDNA synthesis for RT-qPCR

Synonym(s):

cDNA Synthesis Mix

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.55
Pricing and availability is not currently available.

usage

sufficient for 100 reactions
sufficient for 25 reactions
sufficient for 500 reactions

feature

dNTPs included
hotstart: no

technique(s)

RT-PCR: suitable

color

colorless

input

purified RNA

shipped in

dry ice

storage temp.

−20°C

General description

ReadyScript® cDNA Synthesis Mix is a sensitive and easy-to-use solution for two-step RT-PCR. This 5X concentrated mix provides all necessary components (except RNA template) for first-strand synthesis including: buffer, dNTPs, MgCl2, primers, RNase inhibitor protein, ReadyScript reverse transcriptase and stabilizers. ReadyScript is a RNase H(+) derivative of MMLV reverse transcriptase, optimized for reliable cDNA synthesis over a wide dynamic range of input RNA. The unique blend of oligo (dT) and random primers in the ReadyScript cDNA Synthesis Mix works exceptionally well with a wide variety of targets. It is optimized for the production of targets < 1kb in length. ReadyScript cDNA Synthesis Mix produces excellent results in both real-time and conventional RT-PCR.

Application

ReadyScript® cDNA Synthesis Mix has been used for complementary DNA (cDNA) synthesis from total/mRNA[1][2] in 2-step reverse transcription polymerase chain reaction (RT-PCR) and quantitative reverse transcription polymerase chain reaction (RT-qPCR)[3][4] assays for gene expression studies.[5][6]

Features and Benefits

  • Convenient 5X master mix formula provides for quick and easy reaction set up; just add RNA template
  • Includes proprietary blend of randomers and oligodT primers and an RNAse H+ derivative of Moloney murine leukemia virus (MMLV) reverse transcriptase
  • Allows for unbiased, accurate conversion of RNA to cDNA in just 40 minutes
  • Can use up to 1 μg of RNA in a single 20 μL reaction; reactions can also be scaled up
  • Highly stable; can be stored at -20°C for up to 1 year; 4°C for up to one month
  • Specific ratio of randomers and oligo (dT) primers in the mastermix ensures optimal representation of all transcript sequences in the cDNA product
  • High reproducibility: Premixed master mix eliminates variability inherent in other kits requiring mixing of components
  • RNase H(+) Moloney murine leukemia virus (MMLV) reverse transcriptase ensures residual RNA does not interfere with PCR amplification
  • Reverse transcribes both rare and abundant genes with a wide input range of template
  • Convenient 5X master mix formula provides for quick and easy reaction set up; just add RNA template

Components

Supplied as a 5X reaction buffer containing optimized concentrations of MgCl2, dNTPs (dATP, dCTP, dGTP, dTTP), recombinant RNase inhibitor protein, ReadyScript reverse transcriptase, random primers, oligo(dT) primer and stabilizers.

Other Notes

Stable for 1 year when stored in a constant temperature freezer at -20 °C. To extend product′s shelf-life, store the kit at -70 °C. Readyscript cDNA Synthesis Mix showed no loss in functional performance after 20 cycles of freezing on dry ice and thawing on ice. However, we recommend keeping the freeze-thaw cycles to a minimum.

Legal Information

ReadyScript is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extracellular vesicles-derived microRNAs expression as biomarkers for neurological radiation injury: Risk assessment for space exploration.
Gaines D, et al.
Life sciences in space research, 32, 54-62 (2022)
Agnes Weth et al.
Tissue barriers, 5(2), e1285390-e1285390 (2017-04-30)
In the intestine water has to be reabsorbed from the chymus across the intestinal epithelium. The osmolarity within the lumen is subjected to high variations meaning that water transport often has to take place against osmotic gradients. It has been
Water transport through the intestinal epithelial barrier under different osmotic conditions is dependent on LI-cadherin trans-interaction
Agnes Weth
Tissue barriers (2017)
Takashi Kato et al.
Biochemical and biophysical research communications, 549, 83-90 (2021-03-06)
PTEN and p53 are highly mutated in many cancers. These two tumor suppressors have critical functions in the nucleus, such as DNA repair, cell cycle progression, and genome maintenance. However, the in vivo functional relationship of nuclear PTEN and p53 is
Effect of prolonged water stress on essential oil content, compositions and gene expression patterns of mono- and sesquiterpene synthesis in two oregano (Origanum vulgare L.) subspecies.
Morshedloo MR
Plant Physiology and Biochemistry, 111, 119-128 (2017)

Articles

Challenges in gene expression analysis include mRNA stability, temporal transcription patterns, and mRNA-protein correlation, impacting accuracy.

Protocols

Method for reverse transcription of RNA into DNA. Uses a premixed reagent that contains reverse transcriptase, dNTPs, primers, RNase inhibitor and buffer. Fast generation of cDNA.

Reverse transcription analyzes mRNA gene expression, facing challenges like transcript half-life differences and temporal patterns.

Reverse transcription analyzes mRNA gene expression, facing challenges like transcript half-life differences and temporal patterns.

Reverse transcription analyzes mRNA gene expression, facing challenges like transcript half-life differences and temporal patterns.

See All

Related Content

Polymerase chain reaction (PCR) is a technique for amplifying nucleic acid molecules and is commonly used in many applications, including RT-PCR, hot start PCR, end point PCR and more.

Polymerase chain reaction (PCR) is a technique for amplifying nucleic acid molecules and is commonly used in many applications, including RT-PCR, hot start PCR, end point PCR and more.

Polymerase chain reaction (PCR) is a technique for amplifying nucleic acid molecules and is commonly used in many applications, including RT-PCR, hot start PCR, end point PCR and more.

Polymerase chain reaction (PCR) is a technique for amplifying nucleic acid molecules and is commonly used in many applications, including RT-PCR, hot start PCR, end point PCR and more.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service