Skip to Content
Merck

RAB1522

Human IL-22BP ELISA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
41116158
Gene information:
human ... IL22RA2(116379)
Input:
sample type plasma
sample type cell culture supernatant(s)
sample type serum
Species reactivity:
human
Storage temp.:
−20°C
Shipped in:
wet ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1.64 ng/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL22RA2(116379)

General description

This ELISA antibody pair detects human IL-22BP. Other species not yet determined.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Biochem/physiol Actions

Interleukin 22 receptor subunit alpha 2 (IL22RA2) associates with interleukin-22 and prevents it from interacting with other proteins. It may be involved in the regulation of immune responses.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.


pictograms

Corrosion

signalword

Warning

hcodes

Hazard Classifications

Met. Corr. 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library