Skip to Content
Merck

R5876

Rennet from Mucor miehei

Type II

Synonym(s):

Chymosin (primary component)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
EC Number:
232-926-8
NACRES:
NA.61
MDL number:
Biological source:
microbial (Rhizomucor miehei)
Concentration:
≥4% protein (Lowry)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

microbial (Rhizomucor miehei)

Quality Segment

type

Type II

form

powder

concentration

≥4% protein (Lowry)

storage temp.

−20°C

General description

Rennet, an enzyme extract is mainly used in the dairy industry. Chymosin is the main constituent of rennet. In commercial preparation of rennet, other proteases like bovine pepsin is also present in varying concentrations.

Application

Rennet from Mucor miehei has been used:
  • for milk coagulation trials
  • to coagulate the untreated milk and patatin treated milk
  • to coagulate the defatted samples of mammary secretions

Studies have shown that heat treatment of milk causes an increase in the rennet-induced coagulation times. This time was proportional to the temperature of the heat treatment.

Physical form

Powder containing maltodextrin and sodium chloride


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library