Skip to Content
Merck
All Photos(6)

Documents

R4500

Sigma-Aldrich

(−)-Riboflavin

from Eremothecium ashbyii, ≥98%

Synonym(s):

Lactoflavin, Vitamin B2, Vitamin G

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N4O6
CAS Number:
Molecular Weight:
376.36
Beilstein:
97825
EC Number:
MDL number:
UNSPSC Code:
12352205
eCl@ss:
34058005
PubChem Substance ID:
NACRES:
NA.79

biological source

Eremothecium ashbyii

Assay

≥98%

form

powder

color

yellow to orange

mp

290 °C (dec.) (lit.)

storage temp.

room temp

SMILES string

CC1=C(C)C=C(N(C[C@H](O)[C@@H]([C@H](O)CO)O)C(C2=N3)=NC(NC2=O)=O)C3=C1

InChI

1S/C17H20N4O6/c1-7-3-9-10(4-8(7)2)21(5-11(23)14(25)12(24)6-22)15-13(18-9)16(26)20-17(27)19-15/h3-4,11-12,14,22-25H,5-6H2,1-2H3,(H,20,26,27)/t11-,12+,14-/m0/s1

InChI key

AUNGANRZJHBGPY-SCRDCRAPSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Riboflavin belongs to the vitamin-B family and contains a pentose side chain ribitol.

Application

Riboflavin has been used:
  • to check the role of riboflavin diet in rats
  • as a supplement in HECM-6 medium (hamster embryo culture medium)
  • as a supplement in the bacterial basal media

Biochem/physiol Actions

Riboflavin is a key component of the redox cofactors flavin adenine dinucleotide (FAD) and flavin mononucleotide (FMN) that are used by a wide variety of flavoprotein oxidoreductases, dehydrogenase(s), decarboxylase(s) and electron transporters. Riboflavin is involved in various cellular processes, including β oxidation in lipid metabolism, glucose metabolism, protection from cellular oxidative injury, DNA and RNA metabolism, gluconeogenesis, erythropoiesis and generation of corticosteroids. Riboflavin deficiency causes ariboflavinosis with symptoms such as sore throat, hyperemia, edema of oral and mucous membranes, cheilosis and glossitis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The response of young and adult rats to the riboflavin supplementation.
Franca CF
Brazilian Archives of Biology and Technology, 53, 855-860 (2010)
S H McKiernan et al.
Human reproduction (Oxford, England), 15(1), 157-164 (1999-12-28)
The effects of water-soluble vitamins, singly or in combinations, on development of hamster 1-cell embryos were examined in a protein-free, chemically defined culture medium, HECM-6. Pantothenate significantly stimulated blastocyst development compared to the vitamin-free control and to every other single
Stargrove MB, et al.
Herb, Nutrient, and Drug Interactions: Clinical Implications and Therapeutic Strategies (2008)
Ana Rita da Silva Ferreira et al.
Biomolecules, 11(1) (2021-01-02)
Mucositis is a common side-effect of chemotherapy treatment, inducing alterations in the composition of the gut microbiota. Redox active compounds, such as vitamins B2 and C, have been shown to reduce inflammation and enhance the growth of anaerobic bacteria in
Catherine Burgess et al.
Applied and environmental microbiology, 70(10), 5769-5777 (2004-10-07)
This study describes the genetic analysis of the riboflavin (vitamin B(2)) biosynthetic (rib) operon in the lactic acid bacterium Lactococcus lactis subsp. cremoris strain NZ9000. Functional analysis of the genes of the L. lactis rib operon was performed by using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service